Skip to Content
Merck
All Photos(3)

Key Documents

685933

Sigma-Aldrich

Lanthanum-nickel alloy

greener alternative

hydrogen-storage grade

Synonym(s):

LaNi5, Lanthanum pentanickel

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaNi5
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

hydrogen-storage grade

description

Hydrogen Storage Capacity: 1.5-1.6 wt.% @25°C

form

flakes

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[Ni].[La]

InChI

1S/La.Ni

InChI key

DOARWPHSJVUWFT-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Lanthanum-nickel alloy (LaNi5) can be used as a catalyst for the synthesis of multi-walled carbon nanotubes (MWNTs) by chemical vapor deposition (CVD). It can also be used for hydrogen storage applications.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1 - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The electrochemical hydrogen storage of multi-walled carbon nanotubes synthesized by chemical vapor deposition using a lanthanum nickel hydrogen storage alloy as catalyst
Zhang H, et al.
Physica B: Condensed Matter, 352(1-4), 66-72 (2004)
High performance FeTi-3.1 mass% V alloy for on board hydrogen storage solution
Kumar S, et al.
Energy, 75, 520-524 (2014)
Hydrogen storage for carbon nanotubes synthesized by the pyrolysis method using lanthanum nickel alloy as catalyst
Zhang H, et al.
Journal of Applied Physics, 94(10), 6417-6422 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service