Skip to Content
Merck
All Photos(3)

Key Documents

677019

Sigma-Aldrich

(S)-α,α-Bis[3,5-bis(trifluoromethyl)phenyl]-2-pyrrolidinemethanol trimethylsilyl ether

97%

Synonym(s):

(S)-α,α-[3,5-Bis(trifluoromethyl)phenyl]prolinol trimethylsilyl ether, (S)-2-[(Bis(3,5-bis(trifluoromethyl)phenyl)trimethylsilyloxy)methyl]pyrrolidine, (S)-2-[(Bis(3,5-bis(trifluoromethyl)phenyl)trimethylsilanyloxy)methyl]pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23F12NOSi
CAS Number:
Molecular Weight:
597.51
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

48-56 °C

SMILES string

C[Si](C)(C)OC([C@@H]1CCCN1)(c2cc(cc(c2)C(F)(F)F)C(F)(F)F)c3cc(cc(c3)C(F)(F)F)C(F)(F)F

InChI

1S/C24H23F12NOSi/c1-39(2,3)38-20(19-5-4-6-37-19,13-7-15(21(25,26)27)11-16(8-13)22(28,29)30)14-9-17(23(31,32)33)12-18(10-14)24(34,35)36/h7-12,19,37H,4-6H2,1-3H3/t19-/m0/s1

InChI key

MOHRGTBNEJKFMB-IBGZPJMESA-N

General description

(S)-α,α-Bis[3,5-bis(trifluoromethyl)phenyl]-2-pyrrolidinemethanol trimethylsilyl ether is a diarylprolinol silyl ether organocatalyst. It can catalyze a variety of bond-forming reactions such as C-C, C-N, C-O, C-S, and C-Hal in high yields and excellent levels of enantiocontrol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The diarylprolinol silyl ether system: a general organocatalyst.
Jensen KL, et al.
Accounts of Chemical Research, 45(2), 248-264 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service