671401
N-Boc-N′-succinyl-4,7,10-trioxa-1,13-tridecanediamine
95% (HPLC)
Synonym(s):
17-Oxo-6,9,12-trioxa-2,16-diazaeicosanedioic acid 1-(1,1-dimethylethyl)ester
About This Item
Recommended Products
Assay
95% (HPLC)
reaction suitability
reagent type: cross-linking reagent
functional group
Fmoc
amine
carboxylic acid
storage temp.
−20°C
SMILES string
O=C(CCC(O)=O)NCCCOCCOCCOCCCNC(OCC1C2=C(C=CC=C2)C3=C1C=CC=C3)=O
InChI
1S/C19H36N2O8/c1-19(2,3)29-18(25)21-9-5-11-27-13-15-28-14-12-26-10-4-8-20-16(22)6-7-17(23)24/h4-15H2,1-3H3,(H,20,22)(H,21,25)(H,23,24)
InChI key
RTHQDNQOODHVLK-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service