Skip to Content
Merck
All Photos(1)

Key Documents

658340

Sigma-Aldrich

2,4-Dichloro-5-nitropyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4HCl2N3O2
CAS Number:
Molecular Weight:
193.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

0.9 °C/0.5 mmHg

mp

28-32 °C

SMILES string

[O-][N+](=O)c1cnc(Cl)nc1Cl

InChI

1S/C4HCl2N3O2/c5-3-2(9(10)11)1-7-4(6)8-3/h1H

InChI key

INUSQTPGSHFGHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Intermediate in a solid-phase synthesis of diaminopurines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 47, 8897-8897 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service