Skip to Content
Merck
All Photos(3)

Key Documents

590088

Sigma-Aldrich

Sodium metavanadate

anhydrous, 99.9% trace metals basis

Synonym(s):

Sodium (meta)vanadate, Sodium trioxovanadate, Sodium vanadate(V), Sodium vanadium oxide, Sodium vanadium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaVO3
CAS Number:
Molecular Weight:
121.93
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

anhydrous

Assay

99.9% trace metals basis

reaction suitability

core: vanadium
reagent type: catalyst

SMILES string

[Na+].[O-][V](=O)=O

InChI

1S/Na.3O.V/q+1;;;-1;

InChI key

CMZUMMUJMWNLFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Sodium metavanadate has been used as vanadium source in the microbial culture media to study vanadium reduction by mesophilic and thermophilic methanogens.
  • It can be used as a precursor in the preparation of vanadium pentoxide (V2O5) coated carbon nanotubes.
  • It is also used to synthesize corrosion-inhibitive additives for organic coatings.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of metal oxide nanorods using carbon nanotubes as templates.
Satishkumar B C, et al.
Journal of Materials Chemistry, 10(9), 2115-2119 (2000)
Active corrosion protection and corrosion sensing in chromate-free organic coatings.
Buchheit R G, et al.
Progress in Organic Coatings, 47(3-4), 47(3-474) (2003)
Microbial reduction and precipitation of vanadium by mesophilic and thermophilic methanogens.
Zhang J, et al.
Chemical Geology , 370, 29-39 (2014)
Gwendolyn Moise et al.
Biochemistry, 54(42), 6490-6500 (2015-10-08)
Catalysis in protein tyrosine phosphatases (PTPs) involves movement of a protein loop called the WPD loop that brings a conserved aspartic acid into the active site to function as a general acid. Mutation of the tryptophan in the WPD loop
Oluwaseun Mustapha et al.
Environmental toxicology and pharmacology, 38(1), 98-111 (2014-06-14)
Vanadium exposure has been known to lead to lipid peroxidation, demyelination and oligodendrocytes depletion. We investigated behaviour and glial reactions in juvenile mice after early neonatal exposure to vanadium, and examined the direct effects of vanadium in oligodendrocyte progenitor cultures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service