Skip to Content
Merck
All Photos(2)

Key Documents

515426

Sigma-Aldrich

4-Iodophthalonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H3-1,2-(CN)2
CAS Number:
Molecular Weight:
254.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

140-142 °C (lit.)

SMILES string

Ic1ccc(C#N)c(c1)C#N

InChI

1S/C8H3IN2/c9-8-2-1-6(4-10)7(3-8)5-11/h1-3H

InChI key

OOHQHYJZUSXMFD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alkynyl substituted phthalocyanine derivatives as targets for optical limiting
Garcia-Frutos EM, et al.
Journal of Materials Chemistry, 13(4), 749-753 (2003)
The synthesis of a fluorinated phthalocyanine.
Keller TM and Griffith JR.
Journal of Fluorine Chemistry, 13(1), 73-77 (1979)
Vibrational spectra of halophthalonitriles.
Halls MD, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 54(2), 305-317 (1998)
Novel perfluoroalkyl phthalocyanine metal derivatives: Synthesis and photodynamic activities.
Qiu T, et al.
Dyes and Pigments, 83(1), 127-133 (2009)
Canan Uslan et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 24(2), 191-210 (2019-01-24)
Zinc(II) (5), indium(III) (6), and lutetium(III) (7) phthalocyanines (Pcs) peripherally substituted with poly (ethylene glycol) (PEG) monomethyl ether 2000 (PEGME-2000) blocks were synthesized via Sonogashira coupling reaction with high yields and their photophysical, photochemical and photobiological properties were investigated. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service