Skip to Content
Merck
All Photos(1)

Documents

510246

Sigma-Aldrich

Methyl 3,3,3-trifluoropyruvate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COCO2CH3
CAS Number:
Molecular Weight:
156.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.332 (lit.)

bp

86 °C (lit.)

density

1.529 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(=O)C(F)(F)F

InChI

1S/C4H3F3O3/c1-10-3(9)2(8)4(5,6)7/h1H3

InChI key

XGLLQDIWQRQROJ-UHFFFAOYSA-N

Related Categories

General description

Methyl 3,3,3-trifluoropyruvate is an alkyl 3,3,3-trifluoropyruvate that can be synthesized using hexafluoropropene-1,2-oxide (HFPO) as a starting material. It reacts with aromatic amines, benzylic manoamines and diamines to form the corresponding hemiaminals.

Application

Methyl 3,3,3-trifluoropyruvate may be used in the synthesis of:
  • 2-hydroxy-2-trifluoromethylbutan-4-olides
  • 2-(trifluoromethyl)butan-4-olides
  • 4-trifluoromethyl-(2H)-pyridazin-3-ones
  • methyl 3-methoxy-2-trifluoromethylacrylate

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorinated butanolides and butenolides: Part 8. 2-(Trifluoromethyl) butan-4-olides by synthesis from methyl 3, 3, 3-trifluoropyruvate as building block.
Paleta O, et al.
Journal of Fluorine Chemistry, 111(2), 175-184 (2001)
A convenient synthesis of 4-trifluoromethyl-(2H)-pyridazin-3-ones from methyl 3, 3, 3-trifluoropyruvate.
Sibgatulin DA, et al.
Synlett, 12, 1907-1911 (2005)
Methyl 3,3,3-trifluoropyruvate: an improved procedure starting from hexafluoropropene-1, 2-oxide; identification of byproducts.
Dolensky B, et al.
Journal of Fluorine Chemistry, 115(1), 67- 74 (2002)
Methyl 3, 3, 3-trifluoropyruvate hemiaminals: Stability and transaminations.
Dolensky B, et al.
Journal of Fluorine Chemistry, 126(5), 745-751 (2005)
Fluorine-Sacrificial Cyclizations as an Access to 5-Fluoropyrazoles.
Volle JN and Schlosser M.
European Journal of Organic Chemistry, 2000(5), 823-828 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service