Skip to Content
Merck
All Photos(3)

Key Documents

460613

Sigma-Aldrich

Sarcosine tert-butyl ester hydrochloride

97%

Synonym(s):

tert-Butyl sarcosinate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCH2CO2C(CH3)3 · HCl
CAS Number:
Molecular Weight:
181.66
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

137-141 °C (dec.) (lit.)

SMILES string

Cl.CNCC(=O)OC(C)(C)C

InChI

1S/C7H15NO2.ClH/c1-7(2,3)10-6(9)5-8-4;/h8H,5H2,1-4H3;1H

InChI key

RNLQHMIDSCYLAK-UHFFFAOYSA-N

Application

Sarcosine tert-butyl ester hydrochloride may be used for the synthesis of (N-(((tert-butyl)oxycarbonyl)methyl)-N-methylcarbamoyl)methyl(2S)-2-(6-methoxy(2-naphthyl))propanoate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and anti-inflammatory activity of a series of N-substituted naproxen glycolamides: Nitric oxide-donor naproxen prodrugs.
Ranatunge RR, et al.
Bioorganic & Medicinal Chemistry, 14(8), 2589-2589 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service