Skip to Content
Merck
All Photos(1)

Documents

448176

Sigma-Aldrich

1-Hexan-d13-ol

≥98 atom % D, ≥99% (CP)

Synonym(s):

Hexyl-d13 alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)4CD2OH
CAS Number:
Molecular Weight:
115.25
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.13

isotopic purity

≥98 atom % D

Assay

≥99% (CP)

refractive index

n20/D 1.414 (lit.)

bp

156.5 °C (lit.)

mp

−52 °C (lit.)

density

0.918 g/mL at 25 °C

mass shift

M+13

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])O

InChI

1S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2

InChI key

ZSIAUFGUXNUGDI-UTBWLCBWSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuseppe Avellone et al.
Food chemistry, 240, 222-230 (2017-09-28)
The purpose of the present work was to find a correlation between microencapsulation technology applied to wines and the resulting quality of the wine itself in terms of volatile composition and phenolic profile. To this aim, samples of Nero d'Avola
Evangelia Zdrali et al.
ACS nano, 11(12), 12111-12120 (2017-12-12)
Mixtures of nano- and microscopic oil droplets in water have recently been rediscovered as miniature reaction vessels in microfluidic environments and are important constituents of many environmental systems, food, personal care, and medical products. The oil nanodroplet/water interface stabilized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service