Skip to Content
Merck
All Photos(1)

Key Documents

440566

Sigma-Aldrich

Deuteroporphyrin IX dimethyl ester from bovine blood

technical grade, synthetic, 90%

Synonym(s):

Dimethyl 3,7,12,17-tetramethyl-21H,23H-porphine-2,18-dipropionate, NSC 17451

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H34N4O4
CAS Number:
Molecular Weight:
538.64
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

solid

reaction suitability

core: porphyrin
reagent type: catalyst

mp

217-220 °C (lit.)

λmax

399 nm

SMILES string

COC(=O)CCc1c(C)c2cc3[nH]c(cc3C)cc4nc(cc4C)cc5[nH]c(cc1n2)c(CCC(=O)OC)c5C

InChI

1S/C32H34N4O4/c1-17-11-22-14-27-19(3)23(7-9-31(37)39-5)29(35-27)16-30-24(8-10-32(38)40-6)20(4)28(36-30)15-26-18(2)12-21(34-26)13-25(17)33-22/h11-16,34-35H,7-10H2,1-6H3/b21-13-,22-14-,25-13-,26-15-,27-14-,28-15-,29-16-,30-16-

InChI key

JDWKDJYBNJTDOW-RWJCOFQGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of:
  • Deuteroporphyrindimethylester Ru(II) complexes
  • Rh porphyrins
  • Porphyrin-linked nitroimidazole antibiotics
  • Porphyrin-fullerene dyads
  • Porphyrin-platinum complexes
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service