Skip to Content
Merck
All Photos(2)

Documents

410888

Sigma-Aldrich

3-Bromo-3-buten-1-ol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(Br)CH2CH2OH
CAS Number:
Molecular Weight:
151.00
Beilstein:
1737671
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.499 (lit.)

bp

64-65 °C/9 mmHg (lit.)

density

1.522 g/mL at 25 °C (lit.)

SMILES string

OCCC(Br)=C

InChI

1S/C4H7BrO/c1-4(5)2-3-6/h6H,1-3H2

InChI key

RTKMFQOHBDVEBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

3-Bromo-3-buten-1-ol is a primary alcohol. Acetylation of 3-bromo-3-buten-1-ol with acetic acid catalyzed by ytterbium(III) tosylate has been studied.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 27, 3017-3017 (1986)
The Journal of Organic Chemistry, 50, 1621-1621 (1985)
The Journal of Organic Chemistry, 46, 1723-1723 (1981)
Yen-Pin Lu et al.
Biochemical and biophysical research communications, 379(2), 351-355 (2008-12-24)
Octaprenyl diphosphate synthase (OPPs) and undecaprenyl diphosphate synthases (UPPs) catalyze consecutive condensation reactions of farnesyl diphosphate (FPP) with 5 and 8 isopentenyl diphosphate (IPP) to generate C(40) and C(55) products with trans- and cis-double bonds, respectively. In this study, we
Panshin SY.
US Geological Survey Data Series (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service