Skip to Content
Merck
All Photos(1)

Documents

388386

Sigma-Aldrich

4,4′-Methylenebis(cyclohexyl isocyanate), mixture of isomers

90%

Synonym(s):

HMDI, Hydrogenated MDI

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(C6H10NCO)2
CAS Number:
Molecular Weight:
262.35
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

90%

form

liquid

refractive index

n20/D 1.497 (lit.)

mp

26 °C (lit.)

density

1.066 g/mL at 25 °C (lit.)

SMILES string

O=C=NC1CCC(CC1)CC2CCC(CC2)N=C=O

InChI

1S/C15H22N2O2/c18-10-16-14-5-1-12(2-6-14)9-13-3-7-15(8-4-13)17-11-19/h12-15H,1-9H2

InChI key

KORSJDCBLAPZEQ-UHFFFAOYSA-N

General description

4,4′-Methylenebis(cyclohexyl isocyanate), mixture of isomers (HMDI) is an aromatic diisocyanate with good mechanical properties. It is light stable and is resistant to hydrolysis. It is majorly used in the synthesis of polyurethane.

Application

HMDI forms NCO linkages which can be used in the synthesis of polyurethanes for a variety of biomedical applications.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyperbranched polyurethane (HBPU)-urea and HBPU-imide coatings: effect of chain extender and NCO/OH ratio on their properties
Mishra AK, et al.
Progress in Organic Coatings, 74(1), 134-141 (2012)
Effect of different isocyanates on the properties of soy-based polyurethanes
Javni I, et al.
Journal of Applied Polymer Science, 88(13), 2912-2916 (2003)
Electrospinning of L-tyrosine polyurethanes for potential biomedical applications
Shah PN, et al.
Polymer, 50(10), 2281-2289 (2009)
D A Weyel et al.
Toxicology and applied pharmacology, 77(3), 427-433 (1985-03-15)
The use of isocyanates in industry has been increasing and, therefore, the potential for human exposure has also increased. Two such isocyanates are diphenylmethane-4,4'-diisocyanate (MDI) and dicyclohexylmethane-4,4'-diisocyanate (SMDI). Furthermore, there are only a few reports describing the toxicity of these
I R White et al.
Contact dermatitis, 9(4), 300-303 (1983-07-01)
Organic di-isocyanates are used widely in industry. The concomitant respiratory complications are well-known. Cutaneous problems precipitated by skin contact with these chemicals are less well recognised. Allergic contact dermatitis caused by exposure to di-cyclohexylmethane 4,4'-di-isocyanate has been demonstrated in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service