Skip to Content
Merck
All Photos(1)

Key Documents

379921

Sigma-Aldrich

Hydroxylamine hydrochloride

99.995% trace metals basis

Synonym(s):

Hydroxylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2OH · HCl
CAS Number:
Molecular Weight:
69.49
Beilstein:
3539763
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

Assay

99.995% trace metals basis

form

powder, crystals or chunks

pH

2.5-3.5 (20 °C, 50 g/L)

mp

155-157 °C (dec.) (lit.)

density

1.67 g/mL at 25 °C (lit.)

SMILES string

Cl.NO

InChI

1S/ClH.H3NO/c;1-2/h1H;2H,1H2

InChI key

WTDHULULXKLSOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Organosilane amines as potent inhibitors and structural probes of influenza A virus M2 proton channel
  • Lamellarin D analogues as inibitors of topoisomerase I and potential antitumor agents
  • Azapeptide tocolytic agents as inhibitors of prostaglandin F2α receptor for preventing preterm labor
  • Thiazolidinones spiro-fused to indolin-2-ones as potent and selective inhibitors of Mycobacterium tuberculosis protein tyrosine phosphatase B
  • Orally bioavailable quinoline-based antidiabetic dipeptidyl peptidase IV inhibitors targeting Lys554
  • Pyrimidine nucleoside derivatives with nitric oxide donors as antiviral agents
  • Benzyladenosine compounds targeting adenosine A2A receptor and adenosine transporter for neuroprotection
  • Naphthol derivatives as inhibitors of the vanilloid receptor TRPV1 with improved potency in rat cystometry models of urinary incontinence

Biochem/physiol Actions

MAO inhibitor; inhibits platelet aggregation.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

spleen

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ken Berglund et al.
Journal of neuroscience research, 98(3), 422-436 (2019-04-09)
Although molecular tools for controlling neuronal activity by light have vastly expanded, there are still unmet needs which require development and refinement. For example, light delivery into the brain is still a major practical challenge that hinders potential translation of
Klaus Urbahns et al.
Bioorganic & medicinal chemistry letters, 21(11), 3354-3357 (2011-05-03)
We have identified naphthol derivatives as inhibitors of the vanilloid receptor TRPV1 by high throughput screening. The initial lead showed high clearance in rats and has been optimized by enhancing the acidity of the phenol group. Compound 6b has reduced
Hironobu Maezaki et al.
Bioorganic & medicinal chemistry, 19(15), 4482-4498 (2011-07-12)
Dipeptidyl peptidase IV (DPP-4) inhibition is a validated therapeutic option for type 2 diabetes, exhibiting multiple antidiabetic effects with little or no risk of hypoglycemia. In our studies involving non-covalent DPP-4 inhibitors, a novel series of quinoline-based inhibitors were designed
Shi, J.; et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 22, 899-899 (2011)
Salvatore Cananzi et al.
Bioorganic & medicinal chemistry, 19(16), 4971-4984 (2011-07-26)
A novel 5-oxa-6a,8-diazaindeno[2,1-b]phenanthren-7-one scaffold was designed and synthesized as an active analogue of the cytotoxic marine alkaloid Lamellarin D. The design was based on molecular modeling of the site of interaction of Lamellarin D with DNA-topoisomerase I cleavable complex, whereas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service