All Photos(1)
About This Item
Linear Formula:
C6H4-1,2-(CO2K)2
CAS Number:
Molecular Weight:
242.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
solubility
water: soluble 25 mg/mL, clear, colorless
SMILES string
[K+].[K+].[O-]C(=O)c1ccccc1C([O-])=O
InChI
1S/C8H6O4.2K/c9-7(10)5-3-1-2-4-6(5)8(11)12;;/h1-4H,(H,9,10)(H,11,12);;/q;2*+1/p-2
InChI key
GOMCKELMLXHYHH-UHFFFAOYSA-L
General description
Phthalic acid dipotassium salt is the dipotassium salt of 1,2-benzenedicarboxylic acid.
Application
Phthalic acid dipotassium salt may be used in the preparation of brucite-like Ca2+/Al3+ layered double hydroxide (LDH), Ca2Al(OH)6·NO3·2H2O.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Efficient Separation of Terephthalate and Phthalate Anions by Selective Ion-Exchange Intercalation in the Layered Double Hydroxide Ca2Al (OH) 6? NO3? 2H2O.
Millange F, et al.
Chemistry of Materials, 12(7), 1990-1994 (2000)
Szymon Malinowski et al.
Sensors (Basel, Switzerland), 18(12) (2018-11-25)
Development of new, faster methods of biosensor construction is a huge challenge for current science and industry. In this work, biosensor construction was carried out using a new soft plasma polymerization (SPP) method in which a bio-recognition layer of laccase
Henrik Seir Thoke et al.
Journal of biological physics, 44(3), 419-431 (2018-05-26)
We measured temporal oscillations in thermodynamic variables such as temperature, heat flux, and cellular volume in suspensions of non-dividing yeast cells which exhibit temporal glycolytic oscillations. Oscillations in these variables have the same frequency as oscillations in the activity of
Fen Yang et al.
Chemosphere, 93(1), 82-89 (2013-06-08)
This work studies the sorption behaviors of phthalic acid esters (PAEs) on three soils by batch equilibration experiments and quantitative structure property relationship (QSPR) methodology. Firstly, the effects of soil type, dissolved organic matter and pH on the sorption of
Martha M Téllez-Rojo et al.
The Science of the total environment, 461-462, 386-390 (2013-06-12)
Previous studies suggest that prenatal phthalate exposure affects neurodevelopment and behavior during the first years of life. To evaluate the effect of maternal urinary concentrations of phthalate metabolites during pregnancy on mental and psychomotor development in children 24-36 months of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service