Skip to Content
Merck
All Photos(1)

Key Documents

374059

Sigma-Aldrich

3-Bromo-1,1,1-trifluoroacetone

98%

Synonym(s):

1,1,1-Trifluoro-3-bromo-2-propanone, 1,1,1-Trifluoro-3-bromoacetone, 1,1,1-Trifluoro-3-bromopropanone, 1-Bromo-3,3,3-trifluoroacetone, 3-Bromo-1,1,1-trifluoro-2-propanone, Bromomethyl trifluoromethyl ketone, Bromotrifluoro-2-propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COCH2Br
CAS Number:
Molecular Weight:
190.95
Beilstein:
1703387
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.376 (lit.)

bp

87 °C/743 mmHg (lit.)

density

1.839 g/mL at 25 °C (lit.)

functional group

bromo
fluoro
ketone

SMILES string

FC(F)(F)C(=O)CBr

InChI

1S/C3H2BrF3O/c4-1-2(8)3(5,6)7/h1H2

InChI key

ONZQYZKCUHFORE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Bromo-1,1,1-trifluoroacetone reacts with potassium enolate of ethyl 4,4,4-trifluoroacetoacetate to yield ethyl 2,4-bis(trifluoromethyl)-4-hydroxydihydro-3-furoate.

Application

3-Bromo-1,1,1-trifluoroacetone may be used in the synthesis of:
  • 3-nonylthio-1,1,1-trifluoropropan-2-one
  • cyclic tetrapeptides containing trifluoromethyl and pentafluoroethyl ketone as zinc binding functional group
  • perfluoroalkylated trans-allylic alcohols

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Peng et al.
Structure (London, England : 1993), 27(2), 229-240 (2018-12-26)
The N-terminal transactivation domain (NTD) of estrogen receptor alpha, a well-known member of the family of intrinsically disordered proteins, mediates the receptor's transactivation function. However, an accurate molecular dissection of NTD's structure-function relationships remains elusive. Here, we show that the
M Brauer et al.
Biochemistry, 25(8), 2187-2191 (1986-04-22)
G-Actin is a globular protein (Mr 42 300) known to have three cysteine residues that are at least partially exposed and chemically reactive (Cys-10, -284, and -374). When G-actin was reacted with 3-bromo-1,1,1-trifluoropropanone, three resolvable 19F resonances were observed in
Jan H Overbeck et al.
Journal of biomolecular NMR, 74(12), 753-766 (2020-10-01)
Proteins and nucleic acids are highly dynamic bio-molecules that can populate a variety of conformational states. NMR relaxation dispersion (RD) methods are uniquely suited to quantify the associated kinetic and thermodynamic parameters. Here, we present a consistent suite of 19F-based
Binoy Jose et al.
Bioorganic & medicinal chemistry letters, 14(21), 5343-5346 (2004-09-30)
Cyclic tetrapeptides containing trifluoromethyl and pentafluoroethyl ketone as zinc binding functional group were synthesized as potent HDAC inhibitors. Evaluation by human HDAC inhibition assay and p21 promoter assay showed that these inhibitors are promising anticancer agents.
W A McGee et al.
Analytical biochemistry, 189(2), 267-273 (1990-09-01)
A combined NMR and absorbance stopped-flow has been developed for monitoring the kinetics of biochemical reactions. We demonstrate its usefulness in following the alkaline denaturation of human hemoglobin. No glassblowing is required in the fabrication of the apparatus. Commercially available

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service