Skip to Content
Merck
All Photos(1)

Key Documents

343447

Sigma-Aldrich

5-Isopropyl-1,3-cyclohexanedione

99% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H7(=O)2
CAS Number:
Molecular Weight:
154.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99% (GC)

form

solid

mp

63-65 °C (lit.)

functional group

ketone

shipped in

dry ice

SMILES string

[H]O[H].CC(C)C1CC(=O)CC(=O)C1

InChI

1S/C9H14O2.H2O/c1-6(2)7-3-8(10)5-9(11)4-7;/h6-7H,3-5H2,1-2H3;1H2

InChI key

JYJVWRORYGGPHA-UHFFFAOYSA-N

General description

5-Isopropyl-1,3-cyclohexanedione is a cyclic 1,3-diketone. It can be prepared by condensation of isobutylideneacetone with ethyl malonate. 5-Isopropyl-1,3-cyclohexanedione on methylation with methyl iodide yields 5-isopropyl-2-methyl-1,3-cyclohexanedione. A multistep-synthesis of 5-isopropyl-1,3-cyclohexanedione involving aldol condensation, Dieckmann-type annulation, ester hydrolysis and decarboxylation is reported.

Application

5-Isopropyl-1,3-cyclohexanedione may be used as starting reagent for the synthesis of carvotanacetone analogs. It may be used in the preparation of monocyclic terepenes. It may be used in the synthesis of 2H-pyrans, via reaction with α,β-unsaturated aldehydes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monocyclic Terpenes from Cyclic 1, 3-Diketones1.
Frank RL and Hall Jr HK.
Journal of the American Chemical Society, 72(4), 1645-1648 (1950)
A Multistep Synthesis Featuring Classic Carbonyl Chemistry for the Advanced Organic Chemistry Laboratory.
Duff DB, et al.
Journal of Chemical Education, 89(3), 406-408 (2011)
Microwave-Assisted Solvent and Catalyst Free Synthesis of 2H-Pyrans.
Edayadulla N and Lee YR.
Bull. Korean Chem. Soc., 34(10), 2963-2963 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service