Skip to Content
Merck
All Photos(2)

Key Documents

334995

Sigma-Aldrich

Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)

Synonym(s):

Carbonylchlorohydrotris(triphenylphosphine)ruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]3Ru(CO)(Cl)H
CAS Number:
Molecular Weight:
952.40
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: ruthenium
reagent type: catalyst

mp

204-206 °C (dec.) (lit.)

SMILES string

Cl[Ru].[C-]#[O+].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9

InChI

1S/3C18H15P.CO.ClH.Ru/c3*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2;;/h3*1-15H;;1H;/q;;;;;+1/p-1

InChI key

MVJRXTSUBHUUPN-UHFFFAOYSA-M

General description

Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II) is a coordination compound commonly used as a catalyst in organic synthesis reactions such as hydrogenation, transfer hydrogenative C-C bond-forming reactions, transfer hydrogenation, and α-alkylation of ketones with primary alcohols. In addition, it is used as a precursor of pincer ruthenium carbonyl catalysts for hydrogenation.

Application

  • Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II): This ruthenium complex is used as a versatile catalyst for various reactions including hydrogenation, transfer hydrogenation, C-C bond-forming reactions, α-alkylation of ketones with primary alcohols, and as a precursor for pincer ruthenium carbonyl catalysts for hydrogenation. The complex is noted for its physical properties, such as decomposing above its melting point (204-206°C), and its solubility characteristics. It is very insoluble in water, ethanol, and DMSO, sparingly soluble in benzene and acetone, and slightly soluble in chloroform (Kuriyama, 2001).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbonylchlorohydridotris (triphenylphosphine) ruthenium (II)
Kuriyama W
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-7 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service