Skip to Content
Merck
All Photos(2)

Documents

329827

Sigma-Aldrich

Cesium acetate

99.9% trace metals basis

Synonym(s):

Acetic Acid cesium Salt, Cesium Ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCs
CAS Number:
Molecular Weight:
191.95
Beilstein:
3595637
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

solid

mp

194 °C (lit.)

SMILES string

[Cs+].CC([O-])=O

InChI

1S/C2H4O2.Cs/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

ZOAIGCHJWKDIPJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Tuning diketodioxinone reactivity: biomimetic synthesis of the resorcylate antibiotic fungal metabolites ent-W1278A, -B, and -C, using iterative aromatization reactions.: This study demonstrates the use of cesium acetate in tuning diketodioxinone reactivity for the synthesis of antibiotic fungal metabolites, highlighting its role in iterative aromatization reactions (Navarro et al., 2009).
  • Raman spectroscopy and dioxygen adsorption on Cs-loaded zeolite catalysts for butene isomerization.: The research explores the application of cesium acetate in Cs-loaded zeolite catalysts, demonstrating its effectiveness in butene isomerization through Raman spectroscopy and dioxygen adsorption studies (Li & Davis, 2005).
  • Synthesis of the C3-C19 segment of phorboxazole B.: Cesium acetate is utilized in the complex synthesis of the C3-C19 segment of phorboxazole B, showcasing its importance in advanced organic synthesis processes (Vitale et al., 2005).
  • Beneficial effect of cesium salts on Pd-catalyzed hydroxycarbonylation.: This study reveals the advantageous effects of cesium acetate in palladium-catalyzed hydroxycarbonylation reactions, improving reaction efficiency and product yield (Torisawa et al., 2000).

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sundus N Maqbool et al.
British journal of pharmacology, 177(10), 2365-2380 (2020-01-25)
Breast cancer is the leading cause of death in women worldwide, with resistance to current therapeutic strategies, including tamoxifen, causing major clinical challenges and leading to more aggressive and metastatic disease. To address this, novel strategies that can inhibit the
Weikun Lin et al.
ACS applied materials & interfaces, 12(39), 43967-43975 (2020-09-02)
Floating gate transistor photomemory (FGTPM) has been regarded as one of the most prospective nonvolatile photomemory devices because of its compatibility with transistor-based circuits, nondestructive reading, and multilevel storage. Until now, owing to the excellent photoelectric properties, lead-based perovskite nanocrystals
A procedure for alcohol inversion using cesium acetate.
Huffman JW and Desai RC.
Synthetic Communications, 13(7), 553-557 (1983)
Modification of kaolinite surfaces with cesium acetate at 25, 120, and 220 C.
Frost RL, et al.
Langmuir, 15(26), 8787-8794 (1999)
Efficient method for inversion of secondary alcohols by reaction of chloromethanesulfonates with cesium acetate.
Shimizu T, et al.
Tetrahedron Letters, 37(34), 6145-6148 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service