Skip to Content
Merck
All Photos(1)

Key Documents

325511

Sigma-Aldrich

Cerium(III) oxalate hydrate

99.9% trace metals basis

Synonym(s):

Cerium oxalate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce2(C2O4)3 · xH2O
CAS Number:
Molecular Weight:
544.29 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

powder

composition

Degree of hydration, ~9

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤2000 mg/kg Total metallic and rare earth impurities (ICP-MS)

SMILES string

O.O=C(O[Ce]1OC(=O)C(=O)O1)C(=O)O[Ce]2OC(=O)C(=O)O2

InChI

1S/3C2H2O4.2Ce.H2O/c3*3-1(4)2(5)6;;;/h3*(H,3,4)(H,5,6);;;1H2/q;;;2*+3;/p-6

InChI key

WSVMKOQJZBJDJB-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service