All Photos(3)
About This Item
Linear Formula:
(C3H4O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
crosslinking
~0.1 % cross-linked
Quality Level
mol wt
average Mv ~4,000,000
viscosity
40,000-60,000 cP
transition temp
Tg 106 °C
SMILES string
[Na+].[O-]C(=O)C=C
InChI
1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1
InChI key
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1A - Muta. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Laurence Glennon-Alty et al.
Acta biomaterialia, 9(4), 6041-6051 (2012-12-15)
Mesenchymal stem cells (MSCs) can generate chondrocytes in vitro, but typically need to be cultured as aggregates in the presence of transforming growth factor beta (TGF-β), which makes scale-up difficult. Here we investigated if polyacrylate substrates modelled on the functional
Yanxia Wang et al.
International journal of pharmaceutics, 441(1-2), 170-180 (2012-12-15)
A novel galactose-decorated cross-linked micelles (cl-micelles) with ionic cores using cystamine (Cys) as a biodegradable cross-linker was prepared by using block ionomer complexes of poly(ethylene glycol)-b-poly(2-acryloxyethyl-galactose)-b-poly(acrylic acid) (PEG-b-PAEG-b-PAA) and Ca(2+) (PEG-b-PAEG-b-PAA cl-micelles/Cys). Doxorubicin (DOX) was successfully incorporated into the ionic
Imran Khimji et al.
Chemical communications (Cambridge, England), 49(13), 1306-1308 (2013-01-11)
The melting temperature of duplex DNA is much higher in polyanions than in non-ionic polymers with similar ionic strength, suggesting an additional electrostatic contribution on top of the excluded volume effect.
Chunjiao Zhou et al.
Journal of nanoscience and nanotechnology, 13(7), 4627-4633 (2013-08-02)
Poly(acrylic acid) (PAA) coated-Fe3O4 superparamagnetic nano-composites were synthesized through a solvothermal technique by using cheap and environmental friendly iron salts and PAA. Each nano-composite was composed of many small primary nanocrystals. The as-synthesized products were characterized by X-ray diffraction (XRD)
Lei Liu et al.
ACS nano, 7(2), 1368-1378 (2013-01-04)
Hierarchical FeOOH nanostructure array films constructed by different nanosized building blocks can be synthesized at the air-water interface via a bio-inspired gas-liquid diffusion method. In this approach, poly(acrylic acid) (PAA) as a crystal growth modifier plays a crucial role in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service