Skip to Content
Merck
All Photos(2)

Key Documents

305774

Sigma-Aldrich

3,7-Dimethyl-1-octanol

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CH(CH2)3CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
158.28
Beilstein:
1719638
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.4 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥98%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

98-99 °C/9 mmHg (lit.)

density

0.828 g/mL at 20 °C (lit.)

SMILES string

CC(C)CCCC(C)CCO

InChI

1S/C10H22O/c1-9(2)5-4-6-10(3)7-8-11/h9-11H,4-8H2,1-3H3

InChI key

PRNCMAKCNVRZFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,7-Dimethyl-1-octanol is an fragrance ingredient and its toxicologic and dermatologic review as a fragrance ingredient was reported.

Application

3,7-Dimethyl-1-octanol was employed as medium supplement in the anaerobic enrichment cultures of Pseudomonas citronellolis.2

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J Harder et al.
Applied and environmental microbiology, 61(11), 3804-3808 (1995-11-01)
Anaerobic degradation of natural monoterpenes by microorganisms was evaluated by using Pseudomonas citronellolis DSM 50332 and enrichment cultures containing nitrate as an electron acceptor. P. citronellolis grew anaerobically on 3,7-dimethyl-1-octanol and citronellol but not on geraniol, nerol, and alicyclic monoterpenes.
R M Hanif et al.
Chemical & pharmaceutical bulletin, 46(9), 1428-1431 (1998-10-17)
The tetrahydrogeraniol (THG) derivative, ethyl-(3,7-dimethyl octyl thio) acetate (EDOTA) was prepared by reacting tetrahydrogeranyl bromide (obtained by reaction of 40% hydrobromic acid and concentrated sulfuric acid) with ethyl 2-mercaptoacetate, while 3,7-dimethyl octyl propionate (DOP) was synthesized by a common esterification
R M Hanif et al.
Journal of controlled release : official journal of the Controlled Release Society, 55(2-3), 297-302 (1998-10-31)
Poly(acrylic acid) gels containing 5-fluorouracil (5-FU) and tetrahydrogeraniol (THG) were prepared and the effects of THG on 5-FU permeation across the excised rat skin were studied by in vitro methods. Experiments on in vitro permeation of 5-FU across the skin
J Pevsner et al.
The Journal of biological chemistry, 265(11), 6118-6125 (1990-04-15)
We have characterized the odorant binding properties of purified bovine odorant-binding protein (OBP) using as a ligand [3H]3,7-dimethyloctan-1-ol ([3H]DMO). A broad variety of odorants, including terpenes, aldehydes, esters, and musks, bind to OBP with affinities of 0.2 to 100 microM.
A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S139-S141 (2008-07-22)
A toxicologic and dermatologic review of 3,7-dimethyl-1-octanol when used as a fragrance ingredient is presented.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service