Skip to Content
Merck
All Photos(1)

Key Documents

297720

Sigma-Aldrich

2,2,2-Trifluoroethyl acrylate

contains 100 ppm MEHQ as inhibitor, 99%

Synonym(s):

TFEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH2CF3
CAS Number:
Molecular Weight:
154.09
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

contains

100 ppm MEHQ as inhibitor

refractive index

n20/D 1.35 (lit.)

bp

46 °C/125 mmHg (lit.)

density

1.216 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)COC(=O)C=C

InChI

1S/C5H5F3O2/c1-2-4(9)10-3-5(6,7)8/h2H,1,3H2

InChI key

VBHXIMACZBQHPX-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tarek F El-Arabi et al.
Virology journal, 10, 48-48 (2013-02-08)
Comparatively little information is available on members of the Myoviridae infecting low G+C content, Gram-positive host bacteria of the family Firmicutes. While numerous Bacillus phages have been isolated up till now only very few Bacillus cereus phages have been characterized
Julia Kredel et al.
Molecules (Basel, Switzerland), 24(2) (2019-01-20)
The preparation of highly ordered colloidal architectures has attracted significant attention and is a rapidly growing field for various applications, e.g., sensors, absorbers, and membranes. A promising technique for the preparation of elastomeric inverse opal films relies on tailored core/shell
Avinash Kumar et al.
The journal of physical chemistry. A, 124(12), 2335-2351 (2020-03-10)
The photo-oxidation of 2,2,2-trifluoroethyl acrylate (TFEA) (CH2CHC(O)OCH2CF3) initiated by OH radicals and Cl atoms was investigated in tropospheric conditions using both experimental and computational methods. The kinetic measurements were carried out in the temperature range of 268-363 K using the
Dandan Sheng et al.
Journal of materials chemistry. B, 7(31), 4801-4810 (2019-08-08)
Tendons can transmit mechanical force from muscles to bones for movement. However, the mechanical strength of tendons is compromised after surgery, thus causing a high rate of tendon retear. Hence, the design and preparation of biodegradable materials with excellent mechanical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service