Skip to Content
Merck
All Photos(1)

Key Documents

271446

Sigma-Aldrich

Tetraallyltin

97%

Synonym(s):

Tetraallylstannane, Tetraallyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCH2)4Sn
CAS Number:
Molecular Weight:
283.00
Beilstein:
3536269
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

reaction suitability

core: tin

refractive index

n20/D 1.539 (lit.)

bp

69-70 °C/1.5 mmHg (lit.)

density

1.179 g/mL at 25 °C (lit.)

SMILES string

C=CC[Sn](CC=C)(CC=C)CC=C

InChI

1S/4C3H5.Sn/c4*1-3-2;/h4*3H,1-2H2;

InChI key

XJPKDRJZNZMJQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A useful allyl transfer reagent

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service