Skip to Content
Merck
All Photos(1)

Key Documents

254886

Sigma-Aldrich

6,7-Dimethoxycoumarin

98%

Synonym(s):

6,7-Dimethylesculetin, O-Methylisoscopoletin, O-Methylscopoletin, Aesculetin dimethyl ether, Escoparone, Esculetin 6,7-dimethyl ether, Scoparone, Scopoletin methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10O4
CAS Number:
Molecular Weight:
206.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

143-145 °C (lit.)

SMILES string

COc1cc2OC(=O)C=Cc2cc1OC

InChI

1S/C11H10O4/c1-13-9-5-7-3-4-11(12)15-8(7)6-10(9)14-2/h3-6H,1-2H3

InChI key

GUAFOGOEJLSQBT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

6,7-Dimethoxycoumarin inhibits the in vitro growth of Phytophthora citrophthora, Verticillium dahliae, Penicillium digitatum, P. italicum, Colletotrichum gloeosporioides, Diplodia natalensis and Hendersonula toruloidea. Determination and pharmacokinetic study of 6,7-dimethoxycoumarin in rat plasma after intragastric administration of different decoctions of Yinchenhao Tang by reversed-phase HPLC method with UV detection has been reported.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haitao Lv et al.
Journal of separation science, 31(4), 659-666 (2008-02-12)
A simple, sensitive, and validated method was developed for simultaneous determination of scoparone, capillarisin, rhein, and emodin in rat urine by ultra-performance liquid chromatography/electrospray ionization quadruple time-of-flight mass spectrometry (UPLC-MS). The urinary samples were analyzed on an Acquity UPLC BEH
Christopher M Ference et al.
Phytopathology, 110(2), 287-296 (2019-11-13)
Citrus canker (CC), caused by the bacterial pathogen Xanthomonas citri subsp. citri, impacts citrus production in many areas of the world by reducing yields, degrading tree health, and severely blemishing the outer peels of fresh fruit. The relative susceptibility to
Seung Hyo Jung et al.
Journal of the science of food and agriculture, 99(9), 4397-4406 (2019-03-13)
Compounds of the inner shell of chestnut (Castanea crenata) have diverse biological activities, including anti-cancer and anti-oxidant activities. Here we explored the effects of an extract of chestnut inner shells and of its bioactive component scoparone on vascular smooth muscle
Song Yi Lee et al.
Evidence-based complementary and alternative medicine : eCAM, 2018, 3610494-3610494 (2018-09-14)
A cream formulation containing Artemisia capillaris (AC) extract (ACE) was developed for psoriasis therapy. Although ACE can be dissolved in organic solvents, its topical application is restricted because of toxicities. Therefore, a cream formulation was developed for the convenient and
Nadja Soares Vila-Nova et al.
Experimental parasitology, 133(3), 334-338 (2012-12-13)
Leishmaniasis is a zoonotic disease that can manifest itself in visceral and cutaneous form. The aim of this study was to search for new leishmanicidal compounds. Preliminarily, Artemia salina assay was applied to compounds from two plants found in Northeastern

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service