Skip to Content
Merck
All Photos(1)

Key Documents

235350

Sigma-Aldrich

2-Benzylaniline

≥99%

Synonym(s):

2-Aminodiphenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2C6H4NH2
CAS Number:
Molecular Weight:
183.25
Beilstein:
2803264
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

solid

bp

172-173 °C/12 mmHg (lit.)

mp

49-54 °C (lit.)

SMILES string

Nc1ccccc1Cc2ccccc2

InChI

1S/C13H13N/c14-13-9-5-4-8-12(13)10-11-6-2-1-3-7-11/h1-9H,10,14H2

InChI key

DWOBGCPUQNFAFB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Ignacio Andrés et al.
Bioorganic & medicinal chemistry letters, 12(2), 243-248 (2002-01-05)
The synthesis of a series of novel 2-(aminoalkyl)-2,3,3a,8-tetrahydrodibenzo[c,f]isoxazolo[2,3-a]azepine derivatives as well as their 5-HT(2A/2C) and H(1) receptor binding affinities are described. The in vivo activity as potential anxiolytics of the synthesised compounds was measured in a mCPP challenge test. One
New bis (imino) pyridine-iron (II)-and cobalt (II)-based catalysts: synthesis, characterization and activity towards polymerization of ethylene.
Abu-Surrah AS, et al.
Journal of Organometallic Chemistry, 648(1), 55-61 (2002)
Stereoselective synthesis of (R)-(-)-mianserin.
Pawlowska J, et al.
Tetrahedron Asymmetry, 14(21), 3335-3342 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service