Skip to Content
Merck
All Photos(2)

Key Documents

233560

Sigma-Aldrich

4-Hexadecylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15C6H4NH2
CAS Number:
Molecular Weight:
317.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

254-255 °C/15 mmHg (lit.)

mp

53-56 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCc1ccc(N)cc1

InChI

1S/C22H39N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-21-17-19-22(23)20-18-21/h17-20H,2-16,23H2,1H3

InChI key

RBCCQATUVPNPGQ-UHFFFAOYSA-N

Application

4-Hexadecylaniline has been used:
  • in simple solution based method for the gold (Au) metallization of DNA resulting in a Au nanowire network
  • in fabrication of hybrid organized molecular films by Langmuir-Blodgett (LB) technique and their photoluminescent properties has been investigated
  • in seed mediated synthesis of anisotropic rod shaped gold nanoparticles in organic media

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase Transfer of Gold Metallized DNA.
Swami AS, et al.
Journal of Cluster Science, 20(2), 281-290 (2009)
J Freiría-Gándara et al.
Food & function, 9(8), 4429-4442 (2018-08-03)
The antioxidant (AO) efficiencies and the distributions of gallic acid (GA) and a series of alkyl gallates (propyl, PG, butyl, BG, octyl, OG and lauryl, LG) were determined in intact fish oil-in-water emulsions. The efficiency of the AOs in inhibiting
Mafalda Meireles et al.
Journal of the science of food and agriculture, 99(8), 3917-3925 (2019-01-31)
Controlling the interfacial concentrations of antioxidants (AOs) in oil-in-water emulsions can be regarded as a unique approach for increasing the efficiency of AOs in inhibiting the oxidation of lipids. Classical methods to determine the AO distribution in binary systems cannot
Marlene Costa et al.
Journal of the science of food and agriculture, 97(2), 564-571 (2016-04-22)
The relationships between the hydrophilic-lipophilic balance (HLB) of antioxidants (AOs) and their distributions and efficiencies in emulsions are not fully understood. Recent reports indicate that, for series of homologous antioxidants of different hydrophobicity, the variation of their efficiency with the
Emilio A Raimúndez-Rodríguez et al.
Journal of colloid and interface science, 555, 224-233 (2019-08-06)
The distribution of antioxidants (AOs) in O/W emulsions depends on two partition constants, that between oil-interfacial (POI), and that between the aqueous-interfacial (PWI) regions, that need to be determined in the unbroken emulsion to prevent disruptions of the present equilibria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service