Skip to Content
Merck
All Photos(1)

Key Documents

233021

Sigma-Aldrich

4-(Trifluoromethyl)phenylacetic acid

97%

Synonym(s):

(α,α,α-Trifluoro-p-tolyl)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CH2CO2H
CAS Number:
Molecular Weight:
204.15
Beilstein:
2267687
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

82-85 °C (lit.)

SMILES string

OC(=O)Cc1ccc(cc1)C(F)(F)F

InChI

1S/C9H7F3O2/c10-9(11,12)7-3-1-6(2-4-7)5-8(13)14/h1-4H,5H2,(H,13,14)

InChI key

HNORVZDAANCHAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Trifluoromethyl)phenylacetic acid undergoes diolefination mediated by mono-N-protected amino acid ligands in the presence of ethyl acrylate, Pd(OAc)2, KHCO3 and t-amyl alcohol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constructing multiply substituted arenes using sequential palladium(II)-catalyzed C-H olefination.
Keary M Engle et al.
Angewandte Chemie (International ed. in English), 49(35), 6169-6173 (2010-07-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service