Skip to Content
Merck
All Photos(2)

Key Documents

230200

Sigma-Aldrich

Lithium borohydride solution

2.0 M in THF

Synonym(s):

Lithium boron hydride, Lithium hydroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiBH4
CAS Number:
Molecular Weight:
21.78
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

2.0 M in THF

density

0.896 g/mL at 25 °C

SMILES string

[Li+].[H][B-]([H])([H])[H]

InChI

1S/BH4.Li/h1H4;/q-1;+1

InChI key

UUKMSDRCXNLYOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lithium borohydride solution (2M in THF) has been used for the reduction of 5-benzylidene-2,4-thiazolidinediones and 5-benzylidene-4-oxo-2-thiazolidinethiones to form 5-benzyl-2,4-thiazolidinediones and 5-benzyl-4-oxo-2-thiazolidinethiones, respectively.
Reactant for:
  • Preparation of gallium, indium, rhenium and zinc tris(mercaptoimidazolyl)hydroborato complexes
  • Mechano-chemical metathesis reactions
  • Noncatalytic hydrolysis for hydrogen generation
  • Growth of large gold monolayer protected-clusters
  • Anion substitution reactions
  • Dehydrogenation reactions

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regiospecific reduction of 5-benzylidene-2, 4-thiazolidinediones and 4-oxo-2-thiazolidinethiones using lithium borohydride in pyridine and tetrahydrofuran.
Giles RG
Tetrahedron, 56(26), 4531-4537 (2000)
Pascal Martelli et al.
The journal of physical chemistry. A, 114(37), 10117-10121 (2010-09-03)
The hydrogen dynamics in solid and in liquid LiBH4 was studied by means of incoherent quasielastic neutron scattering. Rotational jump diffusion of the BH4- subunits on the picosecond scale was observed in solid LiBH4. The characteristic time constant is significantly
Piter S Miedema et al.
Physical chemistry chemical physics : PCCP, 14(16), 5581-5587 (2012-03-20)
X-Ray Raman Spectroscopy (XRS) is used to study the electronic properties of bulk lithium borohydride (LiBH(4)) and LiBH(4) in porous carbon nano-composites (LiBH(4)/C) during dehydrogenation. The lithium (Li), boron (B) and carbon (C) K-edges are studied and compared with calculations
Prabhat Arya et al.
Journal of combinatorial chemistry, 6(1), 54-64 (2004-01-13)
A diversity-oriented solution and solid-phase synthesis of tetrahydroquinoline-based tricyclic derivatives has been achieved from enantiomerically pure, natural product-like bicyclic scaffold. The solution synthesis of enantiopure bicyclic scaffold was developed by asymmetric hetero Michael reaction. Our approach for the synthesis of
Matthew S Wellons et al.
Nanotechnology, 20(20), 204022-204022 (2009-05-08)
The addition of catalysts to complex hydrides is aimed at enhancing the hydrogen absorption desorption properties. Here we show that the addition of carbon nanostructure C60 to LiBH4 has a remarkable catalytic effect, enhancing the uptake and release of hydrogen.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service