Skip to Content
Merck
All Photos(2)

Key Documents

227897

Sigma-Aldrich

Silver chromate

98%

Synonym(s):

Chromic acid, disilver(I)salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag2CrO4
CAS Number:
Molecular Weight:
331.73
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

density

5.625 g/mL at 25 °C (lit.)

SMILES string

[Ag+].[Ag+].[O-][Cr]([O-])(=O)=O

InChI

1S/2Ag.Cr.4O/q2*+1;;;;2*-1

InChI key

OJKANDGLELGDHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Ox. Sol. 2 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F G Wouterlood et al.
Journal of neuroscience methods, 7(4), 295-308 (1983-04-01)
A procedure is described by which the solid silver chromate precipitate present in Golgi-impregnated neurons is reduced by a photographic developer into metallic silver grains or particles. The remaining silver chromate is subsequently removed, and the fine structure and synaptic
A Angulo et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 42(3), 393-403 (1994-03-01)
We examined the role of chromium reduction in the Golgi-Colonnier method, correlating the quality of neuronal impregnation with the levels of hexavalent (CrVI) and trivalent (CrIII) chromium in the tissue and in the chromation fluid (CF). The concentrations of both
N Fernandez et al.
Journal of the history of the neurosciences, 10(1), 19-26 (2001-07-12)
Knowledge of cerebral structure and function in its modern form can be traced to the neurone doctrine based largely on the work of Santiago Ramón y Cajal [1852-1934] and his lifelong exploitation of the Golgi method. Cajal openly acknowledged his
F G Wouterlood
Stain technology, 61(6), 337-352 (1986-11-01)
Deterioration of Golgi impregnation begins immediately after impregnated tissue blocks are sectioned with the Vibratome. The first signs of deterioration are fading of delicate impregnated processes, the disruption and fragmentation of dendrites, and, eventually, fading of entire neurons. These changes
F G Wouterlood
Microscopy research and technique, 23(4), 275-288 (1992-12-01)
Direct electron microscopy of nervous tissue stained with the Golgi impregnation method is unsatisfactory because the cytoplasm of the cell bodies and processes of the impregnated neurons are completely filled with a compact precipitate of electron dense silver chromate. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service