Skip to Content
Merck
All Photos(2)

Documents

218782

Sigma-Aldrich

Tetraphenylphosphonium bromide

97%

Synonym(s):

Tetraphenylphosphorus bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)4P(Br)
CAS Number:
Molecular Weight:
419.29
Beilstein:
3922383
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

295-300 °C (lit.)

SMILES string

[Br-].c1ccc(cc1)[P+](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20P.BrH/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;/h1-20H;1H/q+1;/p-1

InChI key

BRKFQVAOMSWFDU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Supporting electrolyte for the electroreduction of buckminsterfullerene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Physical Chemistry, 96, 7137-7137 (1992)
Do Kyun Kim et al.
Journal of microbiology and biotechnology, 23(3), 430-435 (2013-03-07)
Multidrug resistance, especially multidrug efflux mechanisms that extrude structurally unrelated cytotoxic compounds from the cell by multidrug transporters, is a serious problem and one of the main reasons for the failure of therapeutic treatment of infections by pathogenic microorganisms as
Yang Zhou et al.
Bioconjugate chemistry, 22(8), 1459-1472 (2011-06-24)
Alteration in mitochondrial transmembrane potential (ΔΨ(m)) is an important characteristic of cancer. The observation that the enhanced negative mitochondrial potential is prevalent in tumor cell phenotype provides a conceptual basis for development of mitochondrion-targeting therapeutic drugs and molecular imaging probes.
Myocardial imaging for mitochondrial membrane potential.
H William Strauss et al.
JACC. Cardiovascular imaging, 5(3), 293-296 (2012-03-17)
Jae Sung Lee et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 28(32), 7945-7953 (2008-08-08)
At the immature calyx of Held, the fast decay phase of a Ca(2+) transient induced by tetanic stimulation (TS) was followed by a period of elevated [Ca(2+)](i) for tens of seconds, referred to as posttetanic residual calcium (Ca(res)). We investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service