Skip to Content
Merck
All Photos(1)

Key Documents

188913

Sigma-Aldrich

Catecholborane

98%

Synonym(s):

Catecholatoborane, Catecholborane (CB), Pyrocatecholborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5BO2
CAS Number:
Molecular Weight:
119.91
Beilstein:
972072
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

reaction suitability

reagent type: reductant

refractive index

n20/D 1.507 (lit.)

bp

50 °C/50 mmHg (lit.)

mp

12 °C (lit.)

density

1.125 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[bH]1oc2ccccc2o1

InChI

1S/C6H5BO2/c1-2-4-6-5(3-1)8-7-9-6/h1-4,7H

InChI key

CENMEJUYOOMFFZ-UHFFFAOYSA-N

Application

A monofunctional hydroborating agent which reduces β-hydroxyketones to 1,3-diols. Effects conjugate reduction of α,β-enones.
Used to prepare B-alkylcatecholboranes which were used, in turn, to generate alkyl radicals forming aryl ethers from quinones. Employed in a preparation of C2-symmetric boron complexes from methylenebis(oxazolines) used for enantioselective reduction of ketones.

Legal Information

Made under U.S. Pat. No. 6,204,405.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

European Journal of Organic Chemistry, 4596-4596 (2006)
Eveline Kumli et al.
Organic letters, 8(25), 5861-5864 (2006-12-01)
Addition of alkyl radicals generated from B-alkylcatecholboranes onto 1,4-benzoquinones leads to substituted hydroquinones in good overall yields. Formation of aryl ethers via a unique radical addition to the oxygen atom of the enone system is the main reaction when bulky
The Journal of Organic Chemistry, 55, 5190-5190 (1990)
The Journal of Organic Chemistry, 55, 5678-5678 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service