Skip to Content
Merck
All Photos(1)

Key Documents

152854

Sigma-Aldrich

4-Methoxybenzyloxycarbonyl azide

95%

Synonym(s):

4-Methoxybenzyl azidoformate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N3CO2CH2C6H4OCH3
CAS Number:
Molecular Weight:
207.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

reaction suitability

reaction type: click chemistry

mp

30-32 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc(COC(=O)N=[N+]=[N-])cc1

InChI

1S/C9H9N3O3/c1-14-8-4-2-7(3-5-8)6-15-9(13)11-12-10/h2-5H,6H2,1H3

InChI key

NQBRFIXRZDTIRQ-UHFFFAOYSA-N

Application

4-Methoxybenzyloxycarbonyl azide was used to prepare N-4-methoxybenzyloxycarbonyl-L-α-aminoadipic acid.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J J Usher et al.
The Biochemical journal, 151(3), 729-739 (1975-12-01)
1. Phenoxymethylpenicillin sulphoxide 4-methoxybenzyl ester was labelled with 3H in its 2-beta-methyl group. Its specific radioactivity was 362 mCi/mmol. 2. Removal of the side chain of this compound yielded the corresponding ester of 6-aminopenicillanic acid sulphoxide and coupling of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service