Skip to Content
Merck
All Photos(1)

Documents

14889

Sigma-Aldrich

2,2′-(4-Methylphenylimino)diethanol

≥97.0% (NT)

Synonym(s):

2,2′-(p-Tolylimino)diethanol, N,N-Bis(2-hydroxyethyl)-p-toluidine, N-(p-Tolyl)diethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4N(CH2CH2OH)2
CAS Number:
Molecular Weight:
195.26
Beilstein:
2099156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (NT)

form

solid

bp

338-340 °C (lit.)

mp

49-53 °C (lit.)

SMILES string

Cc1ccc(cc1)N(CCO)CCO

InChI

1S/C11H17NO2/c1-10-2-4-11(5-3-10)12(6-8-13)7-9-14/h2-5,13-14H,6-9H2,1H3

InChI key

JUVSRZCUMWZBFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2′-(4-Methylphenylimino)diethanol (p-tolyl diethanolamine) was used as an activating amine.

Caution

may discolor to brown on storage

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M P Patel et al.
Biomaterials, 12(7), 649-652 (1991-09-01)
A number of useful room temperature polymerizing resins were formulated, based on poly(ethyl methacrylate) powder and a range of low shrinkage heterocyclic methacrylate monomers. N,N-dimethyl-p-toluidine or p-tolyl diethanolamine were used as activating amines, but the latter material is less active
M Jontell et al.
Journal of dental research, 74(5), 1162-1167 (1995-05-01)
Monomeric resin components from dental composites are toxic to fibroblasts in culture and thus may interfere with the local immune system of the pulp, reducing its effective defense potential, either by cytotoxicity or by a more specific immune mechanism. Therefore
H H Xu et al.
Journal of biomedical materials research, 52(4), 812-818 (2000-10-18)
The strength and toughness of dental core buildup composites in large stress-bearing restorations need to be improved to reduce the incidence of fracture due to stresses from chewing and clenching. The aims of the present study were to develop novel
S Fujisawa et al.
Journal of dental research, 62(7), 803-805 (1983-07-01)
The interaction between phospholipids and stabilizing lysosome agents such as acetylsalicylic acid (ASA), colchicine (CC), prednisolone (PD), etc., was studied using NMR spectroscopy. The peak intensity of protons arising from these compounds was reduced in liposomes containing lecithin and various
K Ikemura et al.
Dental materials journal, 15(2), 132-143 (1996-12-01)
To develop a self-etching primer for ground dentin, 4-acryloxyethyltrimellitic acid (4-AET) was newly synthesized, and the effect of concentrations varying from 0 (control) to 56.5 wt% 4-AET in the water/ HEMA primer on bonding to the dentin was investigated. Bond

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service