Skip to Content
Merck
All Photos(1)

Documents

146617

Sigma-Aldrich

Bathophenanthrolinedisulfonic acid disodium salt hydrate

98%

Synonym(s):

Bathophenanthrolinedisulfonic acid disodium salt trihydrate, 4,7-Diphenyl-1,10-phenanthroline-disulfonic acid disodium salt trihydrate, Disodium bathophenanthrolinedisulfonate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H14N2Na2O6S2 · 3H2O
CAS Number:
Molecular Weight:
590.53
EC Number:
UNSPSC Code:
12352100
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

300 °C

solubility

water: soluble 100 mg/mL, clear to slightly hazy, yellow (to red)

Looking for similar products? Visit Product Comparison Guide

General description

Bathophenanthrolinedisulfonic acid disodium salt forms water soluble complexes with palladium and their catalytic role in oxidation of 2-hexanol has been examined.

Application

Bathophenanthrolinedisulfonic acid disodium salt hydrate was used in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris(dibenzoylmethanate) complex.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intercalation of luminescent Europium (III) complexes in layered double hydroxides.
Sarakha L, et al.
Opt. Mater., 31(3), 562-566 (2009)
D Trinder et al.
The American journal of physiology, 275(2 Pt 1), G279-G286 (1998-08-04)
The mechanisms of uptake of non-transferrin-bound iron by human hepatoma cells (HuH7) were investigated using 59Fe-citrate and [14C]citrate. The amount of iron associated with the cells increased linearly with time, whereas citrate uptake reached a plateau after 45 min, resulting
Sreenivasulu Kilari et al.
Free radical biology & medicine, 48(7), 961-968 (2010-01-26)
Studies in humans and animals have suggested negative interactions of iron and zinc during their intestinal absorption. Further, zinc seems to prevent iron-induced oxidative damage in rats, which was hypothesized to be through the modulation of the intracellular iron signaling
Elizabeth M Zammit et al.
The Analyst, 136(15), 3069-3072 (2011-06-29)
The reaction of a fluorinated iridium complex with cerium(IV) and organic reducing agents generates an intense emission with a significant hypsochromic shift compared to contemporary chemically-initiated luminescence from metal complexes.
A Paolicchi et al.
Journal of investigative medicine : the official publication of the American Federation for Clinical Research, 47(3), 151-160 (1999-04-13)
gamma-Glutamyl transpeptidase (gamma-GT) is found in serum and in the plasma membranes of virtually all cell types. Its physiologic role is to initiate the hydrolysis of extracellular glutathione (GSH), a tripeptide in which cysteine lies between alpha-glycine and gamma-glutamate residues.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service