Skip to Content
Merck
All Photos(1)

Documents

102687

Sigma-Aldrich

Ethylmalonic acid

97%

Synonym(s):

α-Carboxybutyric acid, 2-Ethylpropanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(COOH)2
CAS Number:
Molecular Weight:
132.11
Beilstein:
774334
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

112-114 °C (lit.)

SMILES string

CCC(C(O)=O)C(O)=O

InChI

1S/C5H8O4/c1-2-3(4(6)7)5(8)9/h3H,2H2,1H3,(H,6,7)(H,8,9)

InChI key

UKFXDFUAPNAMPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrícia Fernanda Schuck et al.
Synapse (New York, N.Y.), 67(3), 111-117 (2012-11-20)
Ethylmalonic acid (EMA) accumulates in tissues of patients affected by short-chain acyl-CoA dehydrogenase deficiency and ethylmalonic encephalopathy, illnesses characterized by variable neurological symptoms. In this work, we investigated the in vitro and in vivo EMA effects on Na(+), K(+)-ATPase (NAK)
Patrícia Fernanda Schuck et al.
Neurochemical research, 35(2), 298-305 (2009-09-17)
High concentrations of ethylmalonic acid are found in tissues and biological fluids of patients affected by ethylmalonic encephalopathy, deficiency of short-chain acyl-CoA dehydrogenase activity and other illnesses characterized by developmental delay and neuromuscular symptoms. The pathophysiological mechanisms responsible for the
Carlo Viscomi et al.
Nature medicine, 16(8), 869-871 (2010-07-27)
Ethylmalonic encephalopathy is caused by mutations in ETHE1, a mitochondrial matrix sulfur dioxygenase, leading to failure to detoxify sulfide, a product of intestinal anaerobes and, in trace amounts, tissues. Metronidazole, a bactericide, or N-acetylcysteine, a precursor of sulfide-buffering glutathione, substantially
Maja Di Rocco et al.
Molecular genetics and metabolism, 89(4), 395-397 (2006-07-11)
A child is reported presenting with a clinical picture suggestive of genetic connective tissue disorders (vascular fragility, articular hyperlaxity, delayed motor development, and normal cognitive development), an absence of pathological ethylmalonic acid excretion during inter-critical phases and a homozygous R163W
Imad Dweikat et al.
Metabolic brain disease, 27(4), 613-616 (2012-05-16)
Ethylmalonic encephalopathy (EE) is a rare autosomal recessive disorder caused by mutations in the ETHE1 gene and characterized by chronic diarrhea, encephalopathy, relapsing petechiae and acrocyanosis. Nephrotic syndrome has been described in an infant with EE but the renal histology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service