Microbial Cell Culture
Microbial cell cultures are used in molecular biology for cloning and recombinant protein expression. They are also used in clinical applications to isolate, detect, and identify microbes that cause disease. Microbial culturing enables cell growth and division under controlled laboratory conditions. Bacteria and other microbes can be grown in liquid broth or solid nutrient agar culture media using aseptic techniques to prevent contamination.
Featured Categories
We offer a comprehensive portfolio of antibiotics to prevent contamination of cell cultures through pathogens or other cell lines, for genetic marker selection and cell biology research.
Milli-Q® systems offer innovative water purification technologies engineered to...
Molecular cloning and protein expression methods and protocols for reliable cloning...
Explore microbiological culture media raw materials and supplements, ensuring safety...
Cloning and Recombinant Protein Expression
Bacterial cultures are used for recombinant protein expression, plasmid cloning and amplification, and bacterial artificial chromosome (BAC) and cosmid cloning. Bacterial cells can be altered to uptake and incorporate exogenous genes and plasmids through transformation. Transformation of competent cells typically occurs through electroporation or heat shock of chemically-treated competent cells.
Transformed bacteria are then identified and selected based on acquired antibiotic resistance imparted by recombinant plasmid DNA or using “blue-white” screening to identify recombinant bacteria expressing β-galactosidase as a marker. Yeast can also be transformed using electroporation and other methods for yeast two-hybrid and reporter gene assays to study protein expression and protein interactions.
Cultivation for Microbial Isolation and Identification
Microbial cultures are medically important for the isolation, detection, and differentiation of bacteria and other microbes that cause infectious disease. Characterization and identification using cultivation methods rely on both microbial phenotype and genotype. Cells can be isolated through inoculation of a streak plate to produce pure colonies. Selective media are used to inhibit the growth of certain groups of microbes, allowing others to grow. Differential media contain compounds that enable microbes to be distinguished visually by colony appearance or changes in surrounding media resulting from differences in metabolic or hemolytic activity. These media aid in the identification of bacteria and other microbes through differentiation and elimination.
Looking for More Specific Information?
Visit our document library for data sheets, certificates and technical documentation.
Related Articles
- An introduction to microbial media types, uses and applications. Includes formulations and product selection guide.
- Antifoams are supplied as two basic types of composition or mixtures of the two. Organic antifoams are of synthetic origin. Silicone-based antifoams are generally considered to be siloxane polymers and are also synthetic.
- Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques
- Cell lines for vaccine production and cultivation of SARS CoV-2 and other viruses. Primary, human diploid, and continuous culture cell lines for the propagation of viruses and virus-based vaccines.
- Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.
- See All (16)
Related Protocols
- Technical Article on competent cells. Transformation is a process by which some bacteria take up foreign genetic material (naked DNA) from the environment.
- Initiating a Starter Culture
- General protocols for growth of competent cells in microbial medium.
- The selection of plasmids in yeast is based on the use of auxotrophic mutant strains, which cannot grow without a specific medium component (an amino acid, purine, or pyrimidine)
- Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.
- See All (7)
Find More Articles and Protocols
How Can We Help
In case of any questions, please submit a customer support request
or talk to our customer service team:
Email custserv@sial.com
or call +1 (800) 244-1173
Additional Support
- Chromatogram Search
Use the Chromatogram Search to identify unknown compounds in your sample.
- Calculators & Apps
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- Customer Support Request
Customer support including help with orders, products, accounts, and website technical issues.
- FAQ