MilliporeSigma
HomeQuEChERS Sample Preparation MethodSupel™ QuE Z-Sep Sorbents for QuEChERS
Supel™ QuE Z-Sep Sorbents for QuEChERS
Supel™ QuE Z-Sep sorbents enhance sample cleanup for complex matrices by effectively removing more fat and color from sample extracts than traditional phases for QuEChERS. By eliminating problematic matrix interferences, Z-Sep products provide more robust LC-MS and GC-MS methods. This proprietary technology can replace C18 and PSA phases in your current methods without additional method development.
Supel™ QuE Z-Sep/C18, a combination of Discovery® DSC-18 and Z-Sep particles, is recommended for samples containing <15% fat. Supel QuE Z-Sep+, C18 and Z-Sep dual bonded to silica, is recommended for cleanup of samples containing >15% fat. Supel QuE Z-Sep is recommended for the anlysis of hydrophobic analytes in fatty matrices.
Related products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Rajski Ł, Lozano A, Uclés A, Ferrer C, Fernández-Alba AR. 2013. Determination of pesticide residues in high oil vegetal commodities by using various multi-residue methods and clean-ups followed by liquid chromatography tandem mass spectrometry. Journal of Chromatography A. 1304109-120. https://doi.org/10.1016/j.chroma.2013.06.070
2.
Sapozhnikova Y, Lehotay SJ. 2013. Multi-class, multi-residue analysis of pesticides, polychlorinated biphenyls, polycyclic aromatic hydrocarbons, polybrominated diphenyl ethers and novel flame retardants in fish using fast, low-pressure gas chromatography?tandem mass spectrometry. Analytica Chimica Acta. 75880-92. https://doi.org/10.1016/j.aca.2012.10.034
3.
Geis-Asteggiante L, Lehotay SJ, Lightfield AR, Dutko T, Ng C, Bluhm L. 2012. Ruggedness testing and validation of a practical analytical method for >100 veterinary drug residues in bovine muscle by ultrahigh performance liquid chromatography?tandem mass spectrometry. Journal of Chromatography A. 125843-54. https://doi.org/10.1016/j.chroma.2012.08.020