MilliporeSigma
HomeProtein Expressionβ-Adrenoceptors
β-Adrenoceptors
β-Adrenoceptors are widely distributed, found at both central and peripheral sites, and are activated either via norepinephrine released from sympathetic terminals or via epinephrine (E4250, E4375) released from the adrenal medulla. Important physiological consequences of β-adrenoceptor activation include stimulation of cardiac rate and force, relaxation of vascular, urogenital and bronchial smooth muscle, stimulation of renin secretion from the juxta-glomerular apparatus, stimulation of insulin and glucagon secretion from the endocrine pancreas, stimulation of glycogenolysis in liver and skeletal muscle and stimulation of lipolysis in the adipocyte. Prejunctional β-adrenoceptors are present on some central and peripheral nerve terminals, where their activation results in facilitation of stimulation-evoked neurotransmitter release. However, in contrast to the prejunctional α2-adrenoceptors, these prejunctional receptors do not appear to have major physiologic significance. Most β-adrenoceptor mediated actions involve stimulation of adenylyl cyclase via interaction of the agonist-receptor complex with Gs.
Three β-adrenoceptor proteins have been cloned, and the characteristics of these recombinant receptors correspond with those of the three well characterized β-adrenoceptors on native tissues, designated as β1, β2 and β3. Species differences appear to be important for the β3-adrenoceptor, since several selective β3-adrenoceptor agonists can activate rodent, but not human β3-adrenoceptors. There appear to be multiple affinity states of the β1-adrenoceptor, which may explain the distinct pharmacology of a β-adrenoceptor mediating cardiac contractility.
Many useful pharmacological tools are available for β-adrenoceptor characterization. These include agonists capable of selectively activating β1-, β2- or β3-adrenoceptors, as well as antagonists selective for each of the three subtypes. While it was initially thought that cardiac stimulation involved primarily the β1-adrenoceptor, it now appears that all of the receptor subtypes may be involved. Bronchodilation appears to be mediated primarily by the β2-adrenoceptor. The β3-adrenoceptor is responsible for lipolysis in white adipose tissue and thermogenesis in the brown adipose tissue found in rodents. Renin release appears to be mediated by the β1-adrenoceptor.
β2-Adrenoceptor agonists are commonly used as bronchodilators. Selective β3-adrenoceptor agonists are being developed for the treatment of type II diabetes, obesity and overactive bladder. β-Adrenoceptor antagonists, either non subtype-selective or selective for the β1-adrenoceptor, are widely used as antihypertensives, although the mechanism for this action is still not clearly understood. Intra-ocular administration of nonselective β-adrenoceptor antagonists is a common treatment for glaucoma. Carvedilol, a molecule combining nonselective β-adrenoceptor blockade with α1-adrenoceptor blockade, has recently been shown to produce a dramatic reduction in the mortality/morbidity associated with congestive heart failure.
Currently Accepted Nameβ1 (A6728)β2β3
Alternate Name  atypical β
Structural Information477 aa (human)413 aa (human)408 aa (human)
Subtype Selective AgonistsNorepinephrine (A7257A0937)
Xamoterol
Denopamine (D7815)
T-0509
Procaterol (P9180)
Salbutamol (S8260)
Salbutamol hemisulfate (S5013)
Fenoterol (F1016)
BRL 37344 (B169)
CL 316243 (C5976)
SB-226552
Subtype Selective AntagonistsCGP20712A (C231)
Betaxolol (B5683)
Atenolol (A7655)
ICI-118,551 (I127)
Butoxamine (B1385)
α-Methylpropranolol
SR 58894
SR 59230A (S8688)
Receptor Selective AgonistIsoproterenol (I5627)
Isoproterenol (I5627)
Isoproterenol (I5627)
Receptor Selective AntagonistsAlprenolol
(±)-Propranolol (P0884)
(S)-(–)-Propranolol (P8688)
Pindolol (P0778)
Alprenolol
(±)-Propranolol (P0884)
(S)-(–)-Propranolol (P8688)
Pindolol (P0778)
Bupranolol
Cyanopindolol (C238)
Signal Transduction MechanismsGs (increase cAMP)Gs (increase cAMP)Gs (increase cAMP)
Radioligand of Choice[125I]-I-ICYP[125]-I-ICYP[125I]-I-ICYP
Tissue ExpressionCoronary artery, kidney, heart, CNSKidney, lung, heart, CNSAdipose Tissue, GI tract vascular endothelium
Physiological FunctionCardiac stimulation, coronary vasodilationSmooth muscle relaxationAdipocyte lipolysis, bladder relaxation, thermogenesis
Disease RelevanceHypertension, congestive heart failureAsthmaObesity, diabetes
Abbreviations
BRL 37344: (±)-(R*,R*)-(4-[2-([2-(3-Chlorophenyl)-2-hydroxyethyl]amino)propyl]phenoxy)acedic acid
CGP20712A: (±)-2-Hydroxy-5-[2-[[2-hydroxy-3-[4-[1-methyl-4-(trifluoromethyl)-1H-imidazol-2-yl]phenoxy]propyl]-amino]ethoxy]-benzamide methanesulfonate
CL 316243: (R,R)-5-[2-[[2-(3-Chlorophenyl)-2-hydroxyethyl]-amino]-propyl]1,3-benzodioxole-2,2-dicarboxylate
ICI-118,551: (±)-1-[2,3-(Dihydro-7-methyl-1H-inden-4-yl)oxy]-3-[(1-methylethyl)amino]-2-butanol
ICYP: Iodocyanopindolol
SB-226552: (S)-4-{2-[2-Hydroxy-3-(4-hydroxyphenoxy)propylamino]ethyl}phenoxymethylcyclohexylphosphinic acid
SR 58894: 3-(2-Allylphenoxy)-1-[(1S)-1,2,3,4-tetrahydronaphth-1-ylamino]-(2S)-2-propanol hydrochloride
SR 59230: 3-(2-Ethylphenoxy)-1-[(1S)-1,2,3,4-tetrahydronaphth-1-ylamino]-(2S)-2-propanol oxalate
T-0509: [(–)-(R)-1-(3,4-Dihydroxyphenyl)-2-[(3,4-dimethoxyphenethyl)-amino]ethanol
Similar Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
1.
Girdlestone D. 2000. The IUPHAR compendium of receptor characterization and classification. 2. London: IUPHAR Media.
2.
Collins S. 2001. The  -Adrenergic Receptors and the Control of Adipose Tissue Metabolism and Thermogenesis. Recent Progress in Hormone Research. 56(1):309-328. https://doi.org/10.1210/rp.56.1.309
3.
Evans BA, Sato M, Sarwar M, Hutchinson DS, Summers RJ. 2010. Ligand-directed signalling at ?-adrenoceptors. 159(5):1022-1038. https://doi.org/10.1111/j.1476-5381.2009.00602.x
4.
Zhang Y. 2012. Beta-adrenoceptor signaling pathways mediate cardiac pathological remodeling. Front Biosci. E4(5):1625-1637. https://doi.org/10.2741/e484
5.
Gauthier C, Langin D, Balligand J. 2000. ?3-Adrenoceptors in the cardiovascular system. Trends in Pharmacological Sciences. 21(11):426-431. https://doi.org/10.1016/s0165-6147(00)01562-5
6.
Hieble JP. 2000. Drugs targeting adrenergic receptors: does interaction with a specific subtype confer therapeutic advantage?. Curr Opin Drug Discov Devel. 3(4):370-82.
7.
Hieble JP, Bondinell W, Ruffolo RR. 1995. .alpha.- and .beta.-Adrenoceptors: From the Gene to the Clinic. Part 1. Molecular Biology and Adrenoceptor Subclassification. J. Med. Chem.. 38(18):3415-3444. https://doi.org/10.1021/jm00018a001
8.
Joseph S, Colledge W, Kaumann A. 2004. Aspartate138 is required for the high-affinity ligand binding site but not for the low-affinity binding site of the ?1-adrenoceptor. Naunyn-Schmiedeberg's Arch Pharmacol. 370(3): https://doi.org/10.1007/s00210-004-0962-1
9.
Kolinski M, Plazinska A, Jozwiak K. 2012. Recent Progress in Understanding of Structure, Ligand Interactions and the Mechanism of Activation of the β 2-Adrenergic Receptor. CMC. 19(8):1155-1163. https://doi.org/10.2174/092986712799320547
10.
Ostrowski J, Kjelsberg MA, Caron MG, Lefkowitz RJ. 1992. Mutagenesis of the beta2-Adrenergic Receptor: How Structure Elucidates Function. Annu. Rev. Pharmacol. Toxicol.. 32(1):167-183. https://doi.org/10.1146/annurev.pa.32.040192.001123
11.
Ruffolo RR, Bondinell W, Hieble JP. 1995. .alpha.- and .beta.-Adrenoceptors: From the Gene to the Clinic. 2. Structure-Activity Relationships and Therapeutic Applications. J. Med. Chem.. 38(19):3681-3716. https://doi.org/10.1021/jm00019a001
12.
Sato Y, Kurose H, Isogaya M, Nagao T. 1996. Molecular characterization of pharmacological properties of T-0509 for ?-adrenoceptors. European Journal of Pharmacology. 315(3):363-367. https://doi.org/10.1016/s0014-2999(96)00648-6
13.
Sennitt MV, Kaumann AJ, Molenaar P, Beeley LJ, Young PW, Kelly J, Chapman H, Henson SM, Berge JM, Dean DK, et al. 1998. The contribution of classical (beta1/2-) and atypical beta-adrenoceptors to the stimulation of human white adipocyte lipolysis and right atrial appendage contraction by novel beta3-adrenoceptor agonists of differing selectivities. J Pharmacol Exp Ther. 285(3):1084-95.
14.
Takeda H, Yamazaki Y, Igawa Y, Kaidoh K, Akahane S, Miyata H, Nishizawa O, Akahane M, Andersson K. 2002. Effects of ?3-adrenoceptor stimulation on prostaglandin E2-induced bladder hyperactivity and on the cardiovascular system in conscious rats. Neurourol. Urodyn.. 21(6):558-565. https://doi.org/10.1002/nau.10034
15.
Wehland M, Grosse J, Simonsen U, Infanger M, Bauer J, Grimm D. 2012. The Effects of Newer Beta-Adrenoceptor Antagonists on Vascular Function in Cardiovascular Disease. CVP. 10(3):378-390. https://doi.org/10.2174/157016112799959323
16.
Weyer C, Tataranni PA, Snitker S, Danforth E, Ravussin E. 1998. Increase in insulin action and fat oxidation after treatment with CL 316,243, a highly selective beta3-adrenoceptor agonist in humans. Diabetes. 47(10):1555-1561. https://doi.org/10.2337/diabetes.47.10.1555