MilliporeSigma
HomeProtein ExpressionAdenosine Receptors
Adenosine Receptors
What Are Adenosine Receptors?
Adenosine receptors are members of the superfamily of G protein-coupled receptors and each bears the characteristic motif of seven transmembrane spanning domains. They fall into four known subtypes, referred to as A1, A2A, A2B and A3.
Adenosine Receptors in Disease Research
Modulation of adenosine receptors by selective agonists and antagonists has potential for the treatment of various cardiovascular, inflammatory and neurological diseases. For example, adenosine is released in large amounts during myocardial ischemia and is capable of exerting potent cardioprotective effects in the heart by activating A1 and A3 receptors. A synthetic adenosine receptor agonist might therefore be beneficial to the survival of the ischemic heart. A3 receptor agonists are also a target for cancer therapy. A2A receptor agonists suppress excessive inflammation and reperfusion injury and may be useful also in physiologic stress testing. In the asthmatic lung, however, adenosine acts as an irritant and bronchoconstrictor, such that a synthetic adenosine receptor antagonist would be desirable.
Selectivity for A1 receptors is typically accomplished through modification of the N6-position that gave rise to compounds such as CPA, CHA, and R-PIA, although substantial A3 receptor affinity was later detected for these agonists. The 2-chloro analog CCPA displays slightly greater A1 receptor affinity than the parent CPA. S(-)-ENBA is a highly potent and selective A1 receptor agonist. The affinities of these N6-substituted derivatives at A3 receptors are often intermediate between their respective A1 and A2A affinities. Although most N6-substituted adenosine agonists are A1-selective, the agonist DPMA is 30-fold selective for the rat A2A receptor.
Increased Receptor Potency
Small alkyl amide substitution at the 5'-position, as in NECA, provides increased potency at all of the adenosine receptors. NECA is also among the most potent agonists at all four subtypes of adenosine receptors and is therefore non-selective. CGS21680 is a moderately A2A-selective adenosine agonist in rat but not human, possessing a 140-fold selectivity for A2A versus A1 receptors in rat. Aryl amines related to the agonist CGS21680 (e.g. PAPA-APEC) or the antagonist ZM241,385 can be radioiodinated to provide A2A receptor-selective radioligands.
Selective A3 Adenosine Receptor Agonists and Antagonists
The most recently discovered member of the adenosine receptor family, the A3 receptor, has a unique pharmacological profile, tissue distribution and effector coupling. In recent years, selective A3 adenosine receptor agonists and antagonists have been described. Previously, APNEA had been used as an agent to activate A3 receptors in the presence of non-A3 antagonists, although APNEA is actually 8-fold selective for A1 receptors. IB-MECA is 50-fold selective for A3 versus either A1 or A2A receptors in vitro and appears to be highly A3 selective in vivo. The related 4-amino derivative may be radioiodinated giving rise to [125I]-I-AB-MECA which is widely used as a high affinity radioligand for A3 receptors. 2-Chloro substitution, in combination with modifications at N6 and 5'-positions, e.g. in Cl-IB-MECA (selective for rat A3 vs A1 and A2A receptors by 2500- and 1400-fold, respectively), further enhances A3 selectivity.
Adenosine receptor antagonists, of which xanthines and numerous classes of fused heterocyclic compounds are representative, have been under development as anti-asthmatic, anti-arrhythmic, renal-protective, anti-Parkinson’s and cognition enhancing drugs. A3 receptor antagonists have potential in treating glaucoma or inflammatory disorders.
Xanthine Antagonists
The classical xanthines, theophylline (1,3-dimethylxanthine) and caffeine (1,3,7-trimethylxanthine), are non-selective adenosine receptor antagonists possessing low micromolar affinity. Selective antagonists for A1 receptors include many 8-aryl and 8-cycloalkyl xanthine derivatives, such as CPX which is ~500-fold selective for A1 versus A2A receptors. Certain non-xanthine antagonists, such as N-0841, are A1 selective, whereas SCH-58261 is a highly potent and selective A2A receptor antagonist. CSC and other 8-styrylxanthines, such as KW 6002, are selective for A2A receptors versus both A1 and A2B receptors. However, in dilute solution, these compounds suffer from sensitivity to photoisomerization.
At A3 receptors, the dihydropyridine derivative MRS 1191 (not active at L-type calcium channels) and the triazoloquinazoline MRS 1220 (not selective in rat) are both relatively potent A3 receptor antagonists, possessing Ki values of 31 and 0.65 nM, respectively, at the human subtype. The nucleoside MRS 1292 and the pyridine derivative MRS 1523 are selective A3 adenosine receptor antagonists in the rat and human.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Related Products" section below.
Adenosine Receptor Modulators
Currently Accepted NameA1A2AA2BA3
Alternate NamesRiA2a, RaA2b 
Structural Information326 aa (human)412 aa (human)332 aa (human)318 aa (human)
Receptor Selective AgonistsAdenosine amine congener (A111)
CPA (C8031)
CHA (C9901)
CCPA (C7938)
R-PIA (P4532)
CVT-510
S(-)-ENBA (E111)
CGS21680 (C141)
DPMA
HE-NECAa
ATL-146e
CVT-3146
NECAb (E2387)
IB-MECA (I146)
Cl-IB-MECA (C277)
Receptor Selective AntagonistsCPX (C101)
CPT (C102)
N-0840
WRC-0571
KW 6002
8-(3-Chlorostyryl)caffeine (C197)
SCH-58261 (S4568)
ZM241,385
XACb (X103)
Alloxazine (A28651)
MRS 1754 (M6316)
MRE 2029-F20
I-ABOPXc
MRS 1191 (M227)
MRS 1220 (M228)
MRS 1523 (M1809)
VUF 5574 (V5888)
MRS 1292
MRE 3008F20
Signal Transduction MechanismsGi (cAMP modulation)
increased K+, decreased Ca2+
Gs (increase cAMP)Gs (increase cAMP)
increased Ca2+
Gi (cAMP modulation)
increased Ca2+
Radioligands of Choice[3H]-CHA
[3H]-CPX
[3H]-R-PIA
[3H]-CGS21680
[125I]-PAPA-APEC
[3H]-SCH-58261
[3H]-ZM241,385
[3H]-MRS 1754
[3H]-CPX
[3H]-MRE 2029-F20
[125I]-APNEA
[125I]-AB-MECA
[3H]-PSB-11
Tissue ExpressionCerebral cortex, hippocampus, atria, kidneys, testes, adipocytesPlatelets, endothelial cells, striatum, neutrophilsFibroblasts, colon, aorta, mast cellsEosinophils, heart, lung, pineal
Physiological FunctionInhibition of neurotransmitter release, bradycardia, ischemic preconditioningVasodilatation, inhibition of platelet aggregation, sensorimotor integrationSmooth muscle relaxation, release from mast cellsIschemic preconditioning
Disease RelevanceCardiac arrhythmia, diabetes, strokeParkinson’s disease, inflammation, arthritis, neurodegenerationAsthma, diabetesCardiac ischemia, cancer, arthritis, glaucoma
Footnotes
a) HE-NECA is also a potent agonist at A3 receptors.
b) NECA and XAC are among the most potent agents at this adenosine receptor subtype. However, these compounds are not subtype selective.
c) Rat A3 receptor is relatively insensitive to xanthine blockade.
Abbreviations
AB-MECA: N6-(4-Aminobenzyl)-9-[5-(methylcarbamoyl)-β-D-ribofuranosyl]adenine
ATL-146e: 4-{3-[6-Amino-9-(5-ethylcarbamoyl-3,4-dihydroxy-tetrahydro-furan-2-yl)-9H-purin-2-yl]-prop-2-ynyl}-cyclohexanecarboxylic acid methyl ester
CCPA: 2-Chloro CPA
CGS21680: 2-p-(2-Carboxyethyl)phenethylamino-5’-N-ethylcarboxamidoadenosine
CHA: N6-Cyclohexyladenosine
Cl-IB-MECA: 2-Chloro-N6-(3-iodobenzyl)-9-[5-(methylcarbamoyl)-β-D-ribofuranosyl]adenine
CPA: N6-Cyclopentyladenosine
CPT: 8-Cyclopentyl-1,3-dimethylxanthine
CPX: 8-Cyclopentyl-1,3-dipropylxanthine
CVT-510: N-(3(R)-Tetrahydrofuranyl)-6-aminopurine riboside
CVT-3146: 1-(6-Amino-9-β-D-ribofuranosyl-9H-purin-2-yl)-N-methyl-1H-pyrazole-4-carboxamide (regadenosine)
DBXRM: 1,3-Dibutylxanthine 7-riboside 5'-N-methylcarboxamide
DPMA: N6-[2-(3,5-Dimethoxyphenyl)-2-(2-methylphenyl)ethyl]adenosine
S(-)-ENBA: (2S)-N6-[2-endo-Norbornyl]adenosine
HE-NECA: 2-Hexynyl-adenosine-5’-N-ethyluronamide
I-ABOPX: 3-(3-Iodo-4-aminobenzyl)-8-(4-oxyacetate)-phenyl-1-propyl xanthine
IB-MECA: N6-(3-Iodobenzyl)-9-[5-(methylcarbamoyl)-β-D-ribofuranosyl]adenine
KW 6002: (E)-1,3-Diethyl-8-(3,4-dimethoxyphenylethyl)-7-methyl-3,7-dihydro-1H-purine-2,6-dione
MRE 2029-F20: N-Benzo[1,3]dioxol-5-yl-2-[5-(2,6-dioxo-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purin-8-yl)-1-methyl-1H-pyrazol-3-yloxy]-acetamide
MRE 3008F20: 5N-(4-methoxyphenylcarbamoyl)amino-8-propyl-2-(2-furyl)pyrazolo[4,3-e]-1,2,4-triazolo[1,5-c]pyrimidine
MRS 1191: 3-Ethyl 5-benzyl 2-methyl-6-phenyl-4-phenylethynyl-1,4-(±)-dihydropyridine-3,5-dicarboxylate
MRS 1220: 9-Chloro-2-(2-furyl)-5-phenylacetylamino[1,2,4]-triazolo[1,5-c]quinazoline
MRS 1292: (2R,3R,4S,5S)-2-[N6-3-Iodobenzyl)adenos-9'-yl]-7-aza-1-oxa-6-oxospiro[4.4]-nonan-4,5-diol
MRS 1523: 2,3-Diethyl-4,5-dipropyl-6-phenylpyridine-3-thiocarboxylate-5-carboxylate
MRS 1754: 8-[4-[[(4-Cyano)phenylcarbamoylmethyl]oxy]phenyl]-1,3-di-(n-propyl)xanthine
N-0840: N6-Cyclopentyl-9-methyladenine
NECA: N-Ethylcarboxamidoadenosine
PAPA-APEC: 1-[6-Amino-2-[[2-[4-[3-[[2-[[(4-aminophenyl)acetyl]amino]ethyl]amino]-3-oxopropyl]phenyl]ethyl]amino]-9H-purin-9-yl]-1-deoxy-N-ethyl-β-D-ribofuranuronamide
PIA: R(–)-N6-(2-Phenylisopropyl)adenosine
SCH-58261: 5-Amino-7-(β-phenylethyl)-2-(8-furyl)pyrazolo(4,3-e)-1,2,4-triazolo(1,5-c)pyrimidine
VUF 5574: N-(2-Methoxyphenyl)-N'-(2-(3-pyridyl)quinazolin-4-yl)urea
WRC-0571: 8-(N-Methylisopropyl)amino-N-(5’-endohydroxy-endonorbornyl)-9-methyladenine
XAC: 8-[4-[[[[(2-Aminoethyl)amino]carbonyl]methyl]oxy]phenyl]-1,3-dipropylxanthine; Xanthine amine congener
ZM241,385: 4-[2-(7-Amino-2-(2-furyl)[1,2,4-triazolo[2,3-a] [1,3,5]triazin-5-yl-amino]ethyl phenol
Related Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Armentero MT, Pinna A, Ferré S, Lanciego JL, Müller CE, Franco R. 2011. Past, present and future of A2A adenosine receptor antagonists in the therapy of Parkinson's disease. Pharmacology & Therapeutics. 132(3):280-299. https://doi.org/10.1016/j.pharmthera.2011.07.004
2.
Avila M Y, Stone R A, Civan M M. 2002. Knockout of A3 adenosine receptors reduces mouse intraocular pressure. Investigative ophthalmology & visual science. 43(9):3021-3026.
3.
Baraldi PG, Preti D, Borea PA, Varani K. 2012. Medicinal Chemistry of A3Adenosine Receptor Modulators: Pharmacological Activities and Therapeutic Implications. J. Med. Chem.. 55(12):5676-5703. https://doi.org/10.1021/jm300087j
4.
Baraldi PG, Aghazadeh Tabrizi M, Preti D, Bovero A, Fruttarolo F, Romagnoli R, Moorman AR, Gessi S, Merighi S, Varani K, et al. 2004. [3H]-MRE 2029-F20, a selective antagonist radioligand for the human A2B adenosine receptors. Bioorganic & Medicinal Chemistry Letters. 14(13):3607-3610. https://doi.org/10.1016/j.bmcl.2004.03.084
5.
Baraldi PG, Manfredini S, Simoni D, Zappaterra L, Zocchi C, Dionisotti S, Ongini E. 1994. Synthesis of new pyrazolo[4,3-e]1,2,4-triazolo[1,5-c] pyrimidine and 1,2,3-triazolo[4,5-e]1,2,4-triazolo[1,5-c] pyrimidine displaying potent and selective activity as A2a adenosine receptor antagonists.. Bioorganic & Medicinal Chemistry Letters. 4(21):2539-2544. https://doi.org/10.1016/s0960-894x(01)80279-1
6.
Chen J, Eltzschig HK, Fredholm BB. 2013. Adenosine receptors as drug targets ? what are the challenges?. Nat Rev Drug Discov. 12(4):265-286. https://doi.org/10.1038/nrd3955
7.
Day Y, Huang L, McDuffie MJ, Rosin DL, Ye H, Chen J, Schwarzschild MA, Fink JS, Linden J, Okusa MD. 2003. Renal protection from ischemia mediated by A2A adenosine receptors on bone marrow?derived cells. J. Clin. Invest.. 112(6):883-891. https://doi.org/10.1172/jci15483
8.
Floris M, Sabbadin D, Medda R, Bulfone A, Moro S. 2012. Adenosiland: Walking through adenosine receptors landscape. European Journal of Medicinal Chemistry. 58248-257. https://doi.org/10.1016/j.ejmech.2012.10.022
9.
Fredholm B B, IJzerman A P, Jacobson K A, Klotz K N, Linden J. 2001. International Union of Pharmacology. XXV. Nomenclature and classification of adenosine receptors Pharmacological reviews. 53(4):527-552..
10.
Gao Z, Li Z, Baker S P, Lasley R D, Meyer S, Elzein E, Palle V, Zablocki J A, Blackburn B, Belardinelli L. 2001. Novel short-acting A2A adenosine receptor agonists for coronary vasodilation: inverse relationship between affinity and duration of action of A2A agonists. Journal of Pharmacology and Experimental therapeutics. 298(1):209-218..
11.
Gao Z, Kim S, Biadatti T, Chen W, Lee K, Barak D, Kim SG, Johnson CR, Jacobson KA. 2002. Structural Determinants of A3Adenosine Receptor Activation:  Nucleoside Ligands at the Agonist/Antagonist Boundary. J. Med. Chem.. 45(20):4471-4484. https://doi.org/10.1021/jm020211+
12.
Klotz K. 2000. Adenosine receptors and their ligands. Naunyn-Schmied Arch Pharmacol. 362(4-5):382-391. https://doi.org/10.1007/s002100000315
13.
Kozma E, Suresh Jayasekara P, Squarcialupi L, Paoletta S, Moro S, Federico S, Spalluto G, Jacobson KA. 2013. Fluorescent ligands for adenosine receptors. Bioorganic & Medicinal Chemistry Letters. 23(1):26-36. https://doi.org/10.1016/j.bmcl.2012.10.112
14.
Milne GR, Palmer TM. 2011. Anti-Inflammatory and Immunosuppressive Effects of the A2AAdenosine Receptor. The Scientific World JOURNAL. 11320-339. https://doi.org/10.1100/tsw.2011.22
15.
Muller C. 2000. Adenosine Receptor Ligands-Recent Developments Part I. Agonists. CMC. 7(12):1269-1288. https://doi.org/10.2174/0929867003374101
16.
Ohta A, Sitkovsky M. 2001. Role of G-protein-coupled adenosine receptors in downregulation of inflammation and protection from tissue damage. Nature. 414(6866):916-920. https://doi.org/10.1038/414916a
17.
Salvatore CA, Jacobson MA, Taylor HE, Linden J, Johnson RG. 1993. Molecular cloning and characterization of the human A3 adenosine receptor.. Proceedings of the National Academy of Sciences. 90(21):10365-10369. https://doi.org/10.1073/pnas.90.21.10365
18.
Zhou QY, Li C, Olah ME, Johnson RA, Stiles GL, Civelli O. 1992. Molecular cloning and characterization of an adenosine receptor: the A3 adenosine receptor.. Proceedings of the National Academy of Sciences. 89(16):7432-7436. https://doi.org/10.1073/pnas.89.16.7432