MilliporeSigma
Adenylyl Cyclase
Adenosine 3':5'-monophosphate (cAMP) modifies cell function in all eukaryotic cells, principally through the activation of cAMP-dependent protein kinase (PKA), but also through cAMP-gated ion channels and guanine nucleotide exchange factors directly activated by cAMP. Cellular levels of cAMP reflect the balance between the activities of adenylyl cyclase (ATP: pyrophosphate lyase, cyclizing; EC 4.6.1.1), that catalyzes formation of cAMP from 5'-ATP, and cAMP phosphodiesterases, that catalyze conversion of cAMP to 5'-AMP.
Adenylyl cyclases occur throughout the animal kingdom and play diverse roles in cell regulation. In bacteria, the enzyme may be regulated in response to nutrients or it may constitute a toxic factor in mammals, as with adenylyl cyclases of B. pertussis, B. anthracis, P. aeruginosa, or Y. pestis.
In mammals, the family of adenylyl cyclases is central to one of the most important signal transduction pathways and includes at least ten isozymes. A soluble form (type X) is regulated by HCO3 , whereas the others are membrane-bound and are regulated physiologically through cell-surface receptors linked via heterotrimeric (αβγ) stimulatory (Gαs) and inhibitory (Gαi) guanine nucleotide-dependent regulatory proteins (G-proteins). Most isozymes are activated by Gαs, but differ in their regulation by Gαi and in the effects of Gβγ. These adenylyl cyclases exhibit a putative topology with two tandem repeats of a 6 membrane spanning region and a ~40 kDa cytosolic region. The two cytosolic domains (C1 and C2) share large conserved regions that interact to form a cleft forming the catalytic active site. N-terminal domains are variable and serve regulatory roles. Gαs activates through interaction with the C2 domain yielding the active enzyme: GTP•αs •C. Inhibition by G-proteins occur by a direct effect of Gαi with the C1 domain or by the recombination of βγ with Gαs.
Adenylyl cyclase activity is altered by numerous agents of physiological and biochemical interest. These include agents that act indirectly, by effects on hormone receptors, on Gαs (e.g. cholera toxin) or Gαi (e.g. pertussis toxin), and agents that act directly on the enzyme, in an isozyme-selective manner. All adenylyl cyclases are inhibited by oxidants and are protected by thiols. Most isozymes are stimulated by forskolin. But only select isozymes are activated by Ca2+-calmodulin or regulated by Ca2+ ions. Others are inhibited by nitric oxide (types I and VI) or proteins associated with myc, and several are regulated by protein kinases A and C.
The cleft formed by adenylyl cyclase C1•C2 domains binds both substrate and forskolin. The active site shares topology and reaction mechanism with guanylyl cyclases, with which there is considerable homology, and with oligonucleotide polymerases. Each catalyzes a cation-dependent attack of the 3'-OH on the α-phosphate of an NTP, with PPi as leaving group. Adenylyl cyclases exhibit a reversible bireactant sequential mechanism in which free divalent cation and cation-5'-ATP serve as substrates and cAMP, metal-PPi, and free divalent cation are products.
Although agents that indirectly activate or inhibit adenylyl cyclases are commonly used in the treatment of disease, e.g. β-adrenoceptor blockers, drugs acting directly on the enzyme have only recently been explored. The main classes of which are derivatives of either forskolin or adenine nucleosides. Adenylyl cyclases are inhibited competitively by substrate analogs, the best of which are β-L-2',3'-dd-5’-ATP (IC50 ~24 nM) and, unexpectedly, MANT-5’-GTPγS. Most are also inhibited by adenine nucleoside 3'-polyphosphates, the most potent of which are 2',5'-dd-3’-ATP (IC50~40 nM) and 2’,5’-dd-3’-A4P (IC50 ~7 nM). These latter compounds belong to a class of inhibitors historically called P(purine)-site ligands, which inhibit via a non-competitive, dead-end, post-transition state mechanism. Inhibition by these ligands occurs with varying sensitivity in all isozymes, save those of bacteria and sperm, and they provide an exquisite means for inhibition of this signal transduction pathway.
Cell permeable inhibitors of adenylyl cyclases comprise nucleosides, their derivatives, and recently described pro-nucleotides. The former are effective in the low micromolar range, whereas pro-nucleotides function as prodrugs, with IC50 values in the nanomolar range. These compounds have been used to lower cellular cAMP levels and to alter function in numerous studies with both isolated cells and intact tissues.
Activators
  • Forskolin (F6886) - adenylyl cyclase activator
  • NKH 477 (N3290) - adenylyl cyclase activator (water-soluble)
  • PACAP-27 (A9808) - neuropeptide that stimulates adenylate cyclase
  • PACAP-38 (A1439) - neuropeptide that stimulates adenylate cyclase
  • Adenylyl Cyclase Toxin from Bordetella pertussis (A0847)- converts host ATP to cyclic AMP (cAMP)
Inhibitors
  • NB001 (SML0060) - inhibitor of adenylyl cyclase 1 (AC1)
  • 9-Cyclopentyladenine monomethanesulfonate (C4479) - stable, cell-permeable, non-competitive adenylyl cyclase inhibitor
  • SQ 22,536 (S153) - cell-permeable adenylyl cyclase inhibitor
  • MDL-12,330A hydrochloride (M182)- adenylyl cyclase inhibitor
  • 2′,5′-Dideoxyadenosine (D7408) - cell-permeable adenylyl cyclase inhibitor
  • 2′,5′-Dideoxyadenosine 3′-triphosphate tetrasodium salt (D0939) - potent inhibitor of adenylyl cyclase
  • MANT-GTPγS (M6317) - potent and competitive adenylyl cyclase inhibitor
  • 2′,3′-Dideoxyadenosine (D1285) - specific adenylyl cyclase inhibitor
  • NKY80 (N2165) - selective adenylyl cyclase-V inhibitor
  • KH7 (K3394) - selective inhibitor of soluble adenylyl cyclase
Other Products
  • 1,9-Dideoxyforskolin (D3658) - biologically inactive forskolin analog, useful as a negative control for forskolin
Isozymea
(source)
I
(bovine)
II
(rat)
III
(rat)
IV
(rat)
V
(rat)
Structuresb
amino acids:
1134
1090
1144
1064
1262
accession:M25579M80550M55075M80633M96159
Chromosome7p12 (human)
11A2 (mouse)
5p15 (human)
13C1 (mouse)  
2p22-24 (human)
12A-B (mouse)
14q11.2 (human)
14D3 (mouse)
3q13.2-q21 (human)
16B5 (mouse)
Tissue ExpressioncBrain
Adrenal
Brain
Lungs
Skeletal muscle
Olfactory
Brain
Adrenal
Adipose
Pancreas
UbiquitousHeart
Brain
Physiological FunctiongNeurotransmission
Synaptic plasticity
Memory
Circadian rhythm
Not knownOlfactory response to oderantsNot Known
Cardiac function
Disease RelevanceAdenylyl cyclases mediate signal transduction in all tissues and changes in cellular cAMP levels occur in disease states or may be induced therapeutically in the treatment of disease. Some bacterial toxins raise cAMP levels. Regulation of type V enzyme may be important in cardiomyopathies or of type VI in mitigating aspects of diabetes. Inhibition of adenylyl cyclases may be a useful strategy in combination therapy for treatment of some cancers and for treatment of yeast and a broad range of fungal infections and some cancers.
G-αs
G-αid
G-βhNo change
Forskolin DerivativesRank order of stimulatory potency:
7-deacetyl-forskolin (D3533) ≈ 6-[3-(dimethylamino)propionyl]forskolini ≈ 6-[N-(2-isothiocyanatoethyl)aminocarbonyl]forskolini ≈ 5,6-dehydroxy-7-deacetyl-7-nicotinoylforskolini > 6-acetyl-7-deacetyl-forskolin > 7-deacetyl-7-O-hemisuccinyl-forskolin >> 1,9-dideoxy-forskolin (D3658)i
Ca2+/ CalmodulinNo change(↑↓)No change↓ (no CaM)
PKC/PKA↑ (PKC)↑+++ (PKC)↑ (PKC)↓ (PKC)↑ (PKC) ↓ (PKA)
Competitive InhibitorsjRank-order of inhibitory potency:
β-L-2',3'-dd-5'-ATP ≈ MANT-5’-GTPγS > MANT-5’-ITPγS > β-D-2',3'-dd-5'-ATP ≈ MANT-5’-ATP >> β-L-5'-ATP > 5'-AP(CH2)PPk
Non-competitive Inhibitors
(P-site ligands)k,l
Rank-order of inhibitory potency:
2',5'-dd-3'-A4P > 2'-d-3'-A4P > 2',5'-dd-3'-ATP (D0939) > 2',5'-dd-3'-ADP (D0814) ≥ 2'-d-3'-ATP (D6500) > 2',5'-dd-3'-AMP (D0689) ≥ 2'-d-3'-ADP (D6000) ≥ 2’,5’-dd-2F-Ado > 2',5'-dd-Ado (D7408) > 2'-d-2F-Ado ≥ 9-Xyl-Ade > 2'‑d-3'-AMP > 3'-ADP > 3'-AMP > 2'-d-Ado (D7400) = 3'-d-Ado (C3394) > 5'-d-Ado (D1771) > 9-Ara-Ade (A5762) = 9‑THF-Ade (SQ 22,536) (S153) ≥ 9-CP-Adem (C4479) > Ado (A9251)
Pro-nucleotide Inhibitorsn2’,5’-dd-3’-AMP-bis(Me-SATE) > 2’,5’-dd-3’-AMP-bis(t-Bu-SATE) > 2’,5’-dd-3’-AMP bis(Ph-SATE)
Other InhibitorsRank-order of inhibitory potency:
PMEApp = PMEAp(NH)p > polyadenylate > 2',3'-dd-Ado (D1285) = 2-Cl-2',3'-dd-Ado > PMEA > 2‑amino-7-(2-furanyl)-7,8-dihydro-5(6H)-quinazolinone (NKY80) (N2165) > MDL-12,330A (M182)k
Isozymea
(source)
VI
(rat)
VII
(mouse)
VIII e
(rat)
IX
(mouse)
Xf
(human) (soluble)
Structuresb
amino acids:
1180
1099
1248(A), 1218(B),
1182(C)
1353
1610
accession:M96160U12919L26986U30602AF176813
Chromosome12q12-13 (human)
15F (mouse)
16q12-13 (human)8q24 (human)16p13.3 (human)
16 (mouse)
1q24 (human)
Tissue ExpressioncHeart
Kidney
Brain
Liver
Retina
Brain
Platelets
Brain
Lung
Skeletal muscle
Heart
Brain
Testes
Physiological FunctiongHormonal regulation of gluconeogenesis
Cell proliferation
Ethanol dependencyNeurotransmission
Synaptic plasticity
Memory
Not knownHCO3– sensor
Disease RelevanceAdenylyl cyclases mediate signal transduction in all tissues and changes in cellular cAMP levels occur in disease states or may be induced therapeutically in the treatment of disease. Some bacterial toxins raise cAMP levels. Regulation of type V enzyme may be important in cardiomyopathies or of type VI in mitigating aspects of diabetes. Inhibition of adenylyl cyclases may be a useful strategy in combination therapy for treatment of some cancers and for treatment of yeast and a broad range of fungal infections and some cancers.
G-αsNo change
G-αiNot known
↓  No change
G-βhNot knownNot knownNo change
Forskolin DerivativesRank order of stimulatory potency:
7-deacetyl-forskolin (D3533) ≈ 6-[3-(dimethylamino)propionyl]forskolini ≈ 6-[N-(2-isothiocyanatoethyl)aminocarbonyl]forskolini ≈ 5,6-dehydroxy-7-deacetyl-7-nicotinoylforskolini > 6-acetyl-7-deacetyl-forskolin > 7-deacetyl-7-O-hemisuccinyl-forskolin >> 1,9-dideoxy-forskolin (D3658)i
No changeNo change
Ca2+/ Calmodulin↓ (no CaM)No changeNo changeNot known
PKC/PKA↑(PKC) ↓(PKA)↑(PKC)Not knownNot knownNot known
Competitive InhibitorsjRank-order of inhibitory potency:
β-L-2',3'-dd-5'-ATP ≈ MANT-5’-GTPγS > MANT-5’-ITPγS > β-D-2',3'-dd-5'-ATP ≈ MANT-5’-ATP >> β-L-5'-ATP > 5'-AP(CH2)PPk
Not known
Non-competitive Inhibitors
(P-site ligands)k,l
Rank-order of inhibitory potency:
2',5'-dd-3'-A4P > 2'-d-3'-A4P > 2',5'-dd-3'-ATP (D0939) > 2',5'-dd-3'-ADP (D0814) ≥ 2'-d-3'-ATP (D6500) > 2',5'-dd-3'-AMP (D0689) ≥ 2'-d-3'-ADP (D6000) ≥ 2’,5’-dd-2F-Ado > 2',5'-dd-Ado (D7408) > 2'-d-2F-Ado ≥ 9-Xyl-Ade > 2'‑d-3'-AMP > 3'-ADP > 3'-AMP > 2'-d-Ado (D7400) = 3'-d-Ado (C3394) > 5'-d-Ado (D1771) > 9-Ara-Ade (A5762) = 9‑THF-Ade (SQ 22,536) (S153) ≥ 9-CP-Adem (C4479) > Ado (A9251)
Not known
Pro-nucleotide Inhibitorsn2’,5’-dd-3’-AMP-bis(Me-SATE) > 2’,5’-dd-3’-AMP-bis(t-Bu-SATE) > 2’,5’-dd-3’-AMP bis(Ph-SATE)Not known
Other InhibitorsRank-order of inhibitory potency:
PMEApp = PMEAp(NH)p > polyadenylate > 2',3'-dd-Ado (D1285) = 2-Cl-2',3'-dd-Ado > PMEA > 2‑amino-7-(2-furanyl)-7,8-dihydro-5(6H)-quinazolinone (NKY80) (N2165) > MDL-12,330A (M182)k
Not known
Footnotes
  1. Isozyme source is for structure information.
  2. Structures available at http://www.ncbi.nlm.nih.gov
  3. Tissue expression is for evidence of protein expression; this is not a complete listing.
  4. Empty cells implies no information.
  5. Type VIII exhibits three splice variants (A,B,C).
  6. Type X adenylyl cyclase is stimulated by HCO3.
  7. All adenylyl cyclase isozymes save type X mediate transmembrane regulation of cell function by hormones through altered rates of cAMP formation. The functional consequences of this are thus dependent on tissue-specific receptors and on downstream effectors through which cAMP acts.
  8. Effects of G-βγ are on Gαs-stimulated enzyme.
  9. 6-[3-(dimethylamino)propionyl]-forskolin (NHK477) exhibits enhanced selectivity for type V adenylyl cyclase; 6-[N-(2-isothiocyanatoethyl)aminocarbonyl]forskolin exhibits enhanced selectivity for type II adenylyl cyclase; 5,6-dehydroxy-7-deacetyl-7-nicotinoylforskolin exhibits enhanced selectivity for type III adenylyl cyclase; 1,9-dideoxy-forskolin does not stimulate.
  10. Competitive inhibitors interact with the pre-transition configuration of the enzyme. Isozyme selectivity has not been determined; comparisons are from experiments with a detergent-extract from rat brain.
  11. Inhibitory P-site ligands are characteristically adenine nucleosides or nucleoside phosphates that inhibit adenylyl cyclases by a noncompetitive or uncompetitive, dead-end- (post-transition-state) mechanism. This comparison is from experiments with a detergent-extract from rat brain. These ligands likely inhibit all isozymes, though potencies may vary; comparisons have been made only with types I, II, VI, VII, and VIII.
  12. Of the cell-permeable 9-substituted adenine derivatives, 9-CP-Ade is the most stable chemically and metabolically.
  13. Unprotected nucleotides are not cell-permeable whereas nucleosides, nucleoside analogs, and the protected (pro-) nucleotides are cell permeable. The 3'-bis(acyl-2'-thioethyl)phosphate derivatives of 2',5'-dd-Ado require intact cells for inhibition of cAMP formation and exhibited IC50 values in intact macrophages or adipocytes comparable to the IC50 values seen with the corresponding nucleoside-3'-triphosphate on isolated adenylyl cyclase from these cells. Rank order of inhibitory potency depends on relative rates of uptake, deprotection, and subsequent processing within cells. For this series, deprotection rates are in the order given.
  14. Unprotected nucleotides are not cell-permeable whereas nucleosides, nucleoside analogs, and the protected (pro-) nucleotides are cell permeable. The 3'-bis(acyl-2'-thioethyl)phosphate derivatives of 2',5'-dd-Ado require intact cells for inhibition of cAMP formation and exhibited IC50 values in intact macrophages or adipocytes comparable to the IC50 values seen with the corresponding nucleoside-3'-triphosphate on isolated adenylyl cyclase from these cells. Rank order of inhibitory potency depends on relative rates of uptake, deprotection, and subsequent processing within cells.
For this series, deprotection rates are in the order given.
Abbreviations
Ado: adenosine
2'-d-Ado: 2'-deoxyadenosine
3'-d-Ado: 3'-deoxyadenosine (cordycepin)
5'-d-Ado: 5'-deoxyadenosine
2',5'-dd-Ado: 2',5'-dideoxyadenosine
2',3'-dd-Ado: 2',3'-dideoxyadenosine
9-CP-Ade: 9-(cyclopentyl)-adenine
9-THF-Ade: 9-(tetrahydrofuryl)-adenine (SQ22,536)
9-Ara-Ade: 9-(arabinofuranosyl)-adenine
9-Xyl-Ade: 9-(xylofuranosyl)-adenine
2'-d-3'-AMP: 2'-deoxyadenosine 3'-monophosphate
2'-d-3'-ADP: 2'-deoxyadenosine 3'-diphosphate
2'-d-3'-ATP: 2'-deoxyadenosine 3'-triphosphate
2',5'-dd-3'-AMP: 2',5'-dideoxyadenosine 3'-monophosphate
2',5'-dd-3'-ADP: 2',5'-dideoxyadenosine- 3'-diphosphate
2',5'-dd-3'-ATP: 2',5'-dideoxyadenosine 3'-triphosphate
2',5'-dd-3'-A4P: 2',5'-dideoxyadenosine 3'-tetraphosphate; 5'-APP(CH2)P, adenosine 5'-(βγ-methylene)-triphosphate
βL-5'-ATP: β-L-adenosine 5'-triphosphate; β-L-2',3'-dd-5'-ATP, β-L-2',3'-dideoxyadenosine 5'-triphosphate
PMEA: 9-(2-phosphonylmethoxyethyl)-adenine; PMEApp, 9-(2-diphosphorylphosphonylmethoxyethyl)-adenine
MDL-12,330A: (cis-N-(2-phenylcyclopentyl)azacyclotridec-1-en-2-amine·HCl; NKY80, 2-amino-7-(2-furanyl)-7,8-dihydro-5(6H)-quinazolinone
2',5'-dd-3'-AMP-bis(Me-SATE): 2',5'-dd-Ado-3'-(acetyl-2-thioethyl)-phosphate; 2',5'-dd-3'-AMP-bis(t-Bu-SATE), 2',5'-dd-Ado-3'-(pivaloyl-2-thioethyl)-phosphate
2',5'-dd-3'-AMP-bis(Ph-SATE): 2',5'-dd-Ado-3'-(phenhyl-2-thioethyl)-phosphate; MANT-5'-GTPγS, 3'-(2')-O-N-methylanthraniloyl-guanosine-5'[γ-thio]triphosphate
MANT-5'-ITPγS: 3'-(2')-O-N-methylanthraniloyl-inosine-5'[γ-thio]triphosphate; MANT-5'ATP, 3'-(2')-O-N-methylanthraniloyl-5'-ATP
References
1.
Ahuja N, Kumar P, Bhatnagar R. 2004. The Adenylate Cyclase Toxins. Critical Reviews in Microbiology. 30(3):187-196. https://doi.org/10.1080/10408410490468795
2.
COOPER DMF. 2003. Regulation and organization of adenylyl cyclases and cAMP. 375(3):517-529. https://doi.org/10.1042/bj20031061
3.
Désaubry L, Shoshani I, Johnson RA. 1996. Inhibition of Adenylyl Cyclase by a Family of Newly Synthesized Adenine Nucleoside 3?-Polyphosphates. J. Biol. Chem.. 271(24):14028-14034. https://doi.org/10.1074/jbc.271.24.14028
4.
Dessauer CW, Tesmer JJ, Sprang SR, Gilman AG. 1999. The interactions of adenylate cyclases with P-site inhibitors. Trends in Pharmacological Sciences. 20(5):205-210. https://doi.org/10.1016/s0165-6147(99)01310-3
5.
Dessauer CW. 2009. Adenylyl Cyclase?A-kinase Anchoring Protein Complexes: The Next Dimension in cAMP Signaling. Mol Pharmacol. 76(5):935-941. https://doi.org/10.1124/mol.109.059345
6.
Gille A, Lushington GH, Mou T, Doughty MB, Johnson RA, Seifert R. 2004. Differential Inhibition of Adenylyl Cyclase Isoforms and Soluble Guanylyl Cyclase by Purine and Pyrimidine Nucleotides. J. Biol. Chem.. 279(19):19955-19969. https://doi.org/10.1074/jbc.m312560200
7.
Hanoune J, Defer N. 2001. REGULATION ANDROLE OFADENYLYLCYCLASEISOFORMS. Annu. Rev. Pharmacol. Toxicol.. 41(1):145-174. https://doi.org/10.1146/annurev.pharmtox.41.1.145
8.
Laux WHG, Pande P, Shoshani I, Gao J, Boudou-Vivet V, Gosselin G, Johnson RA. 2004. Pro-nucleotide Inhibitors of Adenylyl Cyclases in Intact Cells. J. Biol. Chem.. 279(14):13317-13332. https://doi.org/10.1074/jbc.m309535200
9.
Pierre S, Eschenhagen T, Geisslinger G, Scholich K. 2009. Capturing adenylyl cyclases as potential drug targets. Nat Rev Drug Discov. 8(4):321-335. https://doi.org/10.1038/nrd2827
10.
Rodbell M. 1971. The glucagon-sensitive adenyl cyclase system in plasma membranes of rat liver. V. An obligatory role of guanyl nucleotides in glucagons action.. J. Biol. Chem.. 2461877-1882 .
11.
Sassone-Corsi P. 2012. The Cyclic AMP Pathway. Cold Spring Harbor Perspectives in Biology. 4(12):a011148-a011148. https://doi.org/10.1101/cshperspect.a011148
12.
Sunahara RK, Dessauer CW, Gilman AG. 1996. Complexity and Diversity of Mammalian Adenylyl Cyclases. Annu. Rev. Pharmacol. Toxicol.. 36(1):461-480. https://doi.org/10.1146/annurev.pa.36.040196.002333
13.
Sutherland E. 1962. Adenyl cyclase: I. Distribution, preparation, and properties.. J. Biol. Chem.. 2371220-1227.
14.
Tesmer JJ. 1997. Crystal Structure of the Catalytic Domains of Adenylyl Cyclase in a Complex with Gs·GTPS. 278(5345):1907-1916. https://doi.org/10.1126/science.278.5345.1907
15.
Zhuo M. 2012. Targeting neuronal adenylyl cyclase for the treatment of chronic pain. Drug Discovery Today. 17(11-12):573-582. https://doi.org/10.1016/j.drudis.2012.01.009
16.
Zippin JH, Levin LR, Buck J. 2001. CO2/HCO3?-responsive soluble adenylyl cyclase as a putative metabolic sensor. Trends in Endocrinology & Metabolism. 12(8):366-370. https://doi.org/10.1016/s1043-2760(01)00454-4