MilliporeSigma
HomeProtein ExpressionGlycolysis via the Embden-Meyerhof-Parnas Glycolytic Pathway
Glycolysis via the Embden-Meyerhof-Parnas Glycolytic Pathway
What is glycolysis?
Glycolysis is the almost universal pathway that converts glucose into pyruvate along with the formation of nicotinamide adenine dinucleotide (NADH) and adenosine triphosphate (ATP). It primarily occurs in the cytoplasm of the cell.
Aerobic and anerobic glycolysis
Under aerobic conditions, the pyruvate passes into the mitochondria where it is completely oxidized by O2 into CO2 and H2O and its chemical energy largely conserved as ATP. Pyruvate generated via aerobic glycolysis feeds into the TCA or Krebs cycle.
In the absence of sufficient oxygen, the pyruvate is reduced by NADH via anaerobic glycolysis or fermentation to a wide range of products, routinely lactate in animals and ethanol in yeasts.
Embden-Meyerhof-Parnas glycolytic pathway
The starting molecule for glycolysis is glucose, a simple and abundant sugar found in carbohydrates, which provides the energy for most cells. Carbohydrates synthesized during photosynthesis act as the main storage molecules of solar energy. When ingested, complex carbohydrates are enzymatically hydrolyzed to monosaccharides, such as starch to D(+)-glucose.
The catabolism of glucose is the primary energy source for short-term requirements.
The remainder of this article will focus on the glycolytic pathway known as the Embden-Meyerhof-Parnas (EMP) pathway, named for its discoverers, Gustav Embden, Otto Meyerhof, and Jakub Karol Parnas. The chemical steps of the pathway are illustrated in the image to the right and via the video below.
Click to download glycolytic pathway chart image.
10 steps in the glycolytic pathway and enzymes of glycolysis
The table below outlines the 10 individual reactions of the glycolytic pathway and the enzyme (with IUBMB Enzyme Commission number) that catalyzes each step.
Glycolysis via the Embden-Meyerhof-Parnas glycolytic pathway
ReactionEnzymeIUBMB EC Number
1. Phosphorylation of glucose. D(+)-Glucose is phosphorylated with ATP to give glucose-6-phosphate.HexokinaseEC 2.7.1.1
2. Isomerization of glucose-6-P to fructose-6-P. The isomerization of glucose-6-phosphate in the second reaction to fructose-6-phosphate occurs via ring-opening and subsequent keto-enol-tautomerization.Glucose-6-phosphate isomeraseEC 5.3.1.9
3. Phosphorylation of fructose-6-P. The third reaction is another phosphorylation with ATP, whereby fructose-6-phosphate is converted to fructose-1,6-bisphosphate.6-P-FructokinaseEC 2.7.1.11
4. Fructose-1,6-bisphosphate to glyceraldehyde phosphate and dihydroxyacetone phosphate. A key branching reaction is the fourth reaction: a ring-opening reaction of fructose-1,6-bisphosphate, which is cleaved in a retro-aldol reaction into D-glyceraldehyde-3-phosphate, and dihydroxyacetone phosphate.Fructose-bisphosphate aldolaseEC 4.1.2.13
5. Isomerization of dihydroxyacetone-P to glyceraldehyde-P. The branch via dihydroxyacetonephosphate is channelled back into D-glyceraldehyde-3-phosphate in the fifth reaction by an isomerization.Triose-phosphate isomeraseEC 5.3.1.9
6. Glyceraldehyde phosphate oxidation & phosphorylation to 1,3-bisphosphoglycerate. In the sixth reaction, the combined D-glyceraldehyde- 3-phosphate from both routes is oxidized at the C1 to a carboxylic acid and then phosphorylated in the 1-position to yield 1,3-bisphospho-D-glycerate.Glyceraldehyde phosphate dehydrogenase
EC 1.2.1.12
7. ATP formation. This phosphate group in the 1-position is transferred in the seventh reaction from the carboxyl group to ADP to give 3-phospho-D-glycerate.Phosphoglycerate kinaseEC 2.7.2.3
8. 3-Phosphoglycerate to 2-phosphoglycerate. The eighth reaction is an isomerization of 3-phospho-D-glycerate to 2-phospho-D-glycerate.Phosphoglycerate mutaseEC 5.4.2.1
9. 2-Phosphoglycerate to phosphonenolpyruvate. The next metabolite, phosphoenolpyruvate, is formed in a dehydration reaction from 2-phospho-D-glycerate.EnolaseEC 4.2.1.11
10. Formation of pyruvate & ATP. The glycolysis pathway from D(+)-glucose to two molecules of pyruvate is concluded by the tenth reaction, which transfers a phosphate group from phosphoenolpyruvate to ADP, thereby giving ATP and pyruvate.Pyruvate kinase
EC 2.7.1.40
Glycolysis animation - video
Enzymes of Glycolysis
2.7.1.1 - Hexokinase
5.3.1.9 - Glucose-6-Phosphase Isomerase
2.7.1.11 - 6-Phosphofructokinase
4.1.2.13 - Fructose-bisphosphate aldolase
1.2.1.12 - Glyceraldehyde-3-phosphate dehydrogenase
2.7.2.3 - Phosphoglycerate kinase
4.2.1.11 - Phosphopyruvate hydratase (enolase)
2.7.1.40 - Pyruvate kinase
1.1.1.27 - Lactate dehydrogenase
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
This article has been expanded from its original form, published in BioFiles 2007, 2.6, 4.