MilliporeSigma
HomeApplicationsMaterials Science and Engineering Contact Lens & Dental Manufacturing
Contact Lens & Dental Manufacturing
Dental milling machine carving out teeth of dental prosthetic form.
Contact lens and dental manufacturing are rapidly evolving disciplines using material and manufacturing innovations to improve ophthalmic and oral health. Partner with us to source quality raw materials and obtain supply chain security from scale-up through production with flexible bulk and customization options. 
Something went wrong, please try again.
Featured Categories
Example of polymer molecular structure
Polymerization Tools
We provide reliable polymerization tools to initiate and manipulate a chemical reaction of monomers to form polymer chains or networks with specific properties for diverse applications.
Shop Products
Petri dish with dynamic M created from high-purity salts
High-Purity Salts
We offer a wide range of high-purity salts, including nitrates, oxalates, halides, sulfates, carbonates, and acetates, available in both anhydrous and hydrated forms. 
Shop Products
High-purity oxides and ceramics used in the production of lightweight, high-performance semiconductors and high-temperature thermal and electrical insulation applications.
Oxides & Ceramics
We offer an extensive range of high purity oxides and ceramics with specialized synthesis and purification techniques, and distinct particle sizes.
Shop Products
Computer illustration of a spherical gold nanoparticle floating through abstract background.
Inorganic & Metallic Nanomaterials
We offer a comprehensive portfolio of inorganic and metallic nanomaterials, functionalized nanoparticles, and nanomaterial kits for your research needs.
Shop Products
Contact lens with sensor in middle held on fingertip.
Dental prosthetic device fabrication
Contact Lens Manufacturing
Contact lenses are ophthalmic prosthetic devices used for vision correction, therapeutics, and cosmetic applications. Contact lens are typically produced from polymer- or silicone-hydrogel materials due to their enhanced oxygen and water permeability. Material selection also influences surface smoothness, UV radiation absorption, and resistance to tearing and surface defects. Precision manufacturing technologies have enabled further tailoring of contact lens properties, including molding processes, lathe cutting, and plasma processing. To alleviate adverse effects from infection and inflammation and improve consumer comfort and safety, research is currently focused on developing new monomers, polymer chemistries, and manufacturing methods.  
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents
Dental milling machine carving out teeth of dental prosthetic form.
Dental Manufacturing
Dental medicine aims to manage oral health, combat and treat oral disease, and correct teeth and jaw misalignment. Dental manufacturing utilizes diverse solid-state and polymeric syntheses, processing, and surface functionalization methods to fabricate materials, products, and equipment used in dentistry. Fillings, caps, crowns, and bridges are made from resin-based composites; glass ionomers and alloys are used for restorative dentistry applications. Dental impressions use alginate materials to form accurate negative molds for 3D models of teeth and soft tissues. Aesthetic dental care applications include teeth bleaching with peroxide compounds and porcelain veneer bonding to teeth with light-cured resin cements.
Research in dental materials is focused on next-generation monomers to enhance flexibility, durability, biocompatibility, and antimicrobial properties. Utilizing computer-aided design (CAD) and computer-aided manufacturing (CAM), custom-fitted dentures, dental implants, and orthodontic devices, can now be made quickly and inexpensively using 3D printing.  