MilliporeSigma
HomeWet Chemical AnalysisMilli-Q® IQ 7003/05/10/15 Ultrapure & Pure Lab Water Purification System
Milli-Q® IQ 7003/05/10/15 Ultrapure & Pure Lab Water Purification System
Superior Quality Type 1 & Type 2 Water Purification from Tap
See for yourself how the Milli-Q® IQ 7003/05/10/15 ultrapure and pure lab water purification system is designed to exceed your most demanding expectations – from improved productivity and versatility to reduced environmental impact.
This fully-integrated water purification system is your ultimate lab water solution providing: 
  • Superior and consistent quality Type 1 and Type 2 water directly from tap water
  • No maintenance or hazardous chemicals with Elix® EDI technology
  • Intuitive touchscreens
  • Ergonomic Q-POD® and E-POD® dispensers
  • Greater confidence in lab productivity with MyMilli-Q™ digital services
  • A reduced environmental footprint with the mercury-free UV lamps and less electricity, materials and packaging  
Milli-Q IQ 7003/05/10/15 systems awarded Sustainable Lab Product of 2022
Milli-Q® IQ 7003/05/10/15 systems were voted “Sustainable Lab Product of the Year” in the SelectScience® Scientists’ Choice Awards 2022.
Simple & Intuitive Lab Water Dispensing
You’ll always have convenient and easy access to purified water with our ergonomic Q-POD® ultrapure and E-POD® pure water dispensers. Just twist the dispense wheel or tap the touchscreen menu to access a range of dispensing functions.
Consistently Superior Quality Purified Water
Our exclusive, advanced and sustainable purification technologies—including mercury-free ech2o®  UV lamps, Elix® EDI technology and IQnano™ ion-exchange resin—deliver highest-quality Type 1 and Type 2 water for your critical experiments. Our range of POD-Paks lets you further match water quality to your every application. And with accurate, rapid and continuous resistivity and TOC monitoring directly on the POD touchscreens, you’ll always have full confidence in your water quality.
Assured Protection of Stored Pure Water
Our redesigned intelligent pure water storage solution ensures that high-quality Type 2 water is always on hand and ready to use. Unique design features protect against stagnation and bacterial contamination, safeguarding the quality of stored pure water better than ever before.
Unparalleled Ease of Use
Choose a dispense option, instantly monitor water quality parameters, or rapidly retrieve data. Easy-to-navigate touchscreens on each POD dispenser give you quick access and control of essential system functions in just a few clicks.
Space-saving & Clutter-free
Save valuable space as only the slim POD dispenser stays on the bench. The compact purification unit and tank can be tucked away under the bench, on the wall, or a combination of both to best fit your lab’s space. A range of production flow rates (3,5,10,15 L/h) and tank size (25,50,100L) options are available to ensure your lab's needs can be met today and tomorrow. For labs needing more than one point-of-use, up to 4 dispensers can be connected to one system (1 E-POD® and 3 Q-POD® units, or 4 Q-POD® units).
Effortless Data Management
Simplified data management is an integral part of the Milli-Q® IQ 7003/05/10/15 system. In a few clicks, download an individual Dispense Report, determine water quality over a specified time frame, allocate costs for shared resources, or create a customized report. Traceability is also ensured through the e-Sure tags on all consumables.
Completely Mercury Free, Water & Energy Savings, and a Smaller Footprint
  • The first and only mercury-free tap-to-ultrapure water system, with patented built-in mercury-free ech2o® UV lamps
  • Reduced water consumption with the Advanced RO water recovery loop and periodic EDI rinsing
  • Up to 28% reduced system electricity consumption thanks to optimization of components and processes as well as an improved Lab Close mode*
  • Reduced size of systems, tanks and consumables*
  • New, patented Drain Cap purges ~50% of water from the cartridges, reducing the cartridges’ total weight by ~25% before disposal
  • Greener packaging* includes 100% recycled cardboard with sustainable forestry certification and bio-based PE for protective inserts
*Comparisons are versus the previous generation of systems or cartridges
Greener Alternative Products
Milli-Q® IQ 7003/05/10/15 systems and purification kits have been evaluated on 7 dimensions that have a critical impact on Global Warming Energy & Emissions, Water, Packaging, Materials, Suppliers & Manufacturing, CircularEconomy, and Usability & Innovation.
Identified as Greener Alternative Products, we certify that these systems use less electricity, materials and packaging, and are completely mercury free. View the Design for Sustainability (DfS) scorecards for each system:
Greener Alternative Products Icon
Quality & Compliance
Validated and qualified design, combined with our strict manufacturing Quality Assurance system, is your best warranty. We offer a complete range of services, including validation support, which is provided by certified field service engineers using calibrated equipment and a Milli-Q®-specific Qualification Workbook. This Milli-Q® IQ series of systems is intended to produce pure and ultrapure water that meets or exceeds requirements as described by the ASTM, ISO 3696, CLSI, European Pharmacopoeia and US Pharmacopeia.
Digital Services Simplify System Care and Assure Productivity
MyMilli-Q™ Remote Care is a unique online monitoring and service feature that provides:
  • Remote access to your system and its data 
  • Automatic notifications, by email or SMS, in case of an alert or alarm
  • Faster service with remote diagnostics and repair possible
  • Simplified data management and advanced traceability that eases audit and accreditation processes
MyMilli-Q™ Digital Services Sticker Icon
View the Convenience of Connectivity video on how a connected water purification system can support your uninterrupted lab productivity.
Lab Water Applications
Ultrapure Water Applications
  • Chromatography: HPLC, LC-MS, UHPLC, ion chromatography (IC)
  • Elemental analyses: AAS, ICP-MS, ICP-OES
  • Life science applications: Cell culture, PCR, sequencing, DNA microarrays, protein analyses
Pure Water Applications
  • Analytical techniques with sensitivity at ppm level or above, such as pH measurements, UV spectrophotometry, Kjeldahl testing
  • Reagent preparation for dissolution testing, chemical synthesis, microbiology media preparation
  • Staining solutions preparation for histology
  • Feed for instruments: glassware washers, stainless steel autoclaves, stability and weathering test chambers, clinical analyzers, hydrogen generators
Water Specifications / International Conformity
1 These values are typical and may vary depending on the nature and concentration of contaminants in the feed water.
2 Resistivity can also be displayed non-temperature-compensated as required by USP.
3 In the appropriate operating conditions, otherwise typically ≤ 5 ppb.
4 With Millipak® filter or Biopak® polisher.
5 With Millipak® filter or Biopak® polisher when installed and used in a laminar flow hood.
6 With Biopak® polisher when installed and used in a laminar flow hood.
7 With Biopak® polisher.
Feed water requirements
International Regulation Requirements
Declaration of CE and cUL Conformity:
The Milli-Q® IQ 7003, Milli-Q® IQ 7005, Milli-Q® IQ 7010 and Milli-Q® IQ 7015 systems have been tested by an independent and accredited company for compliance with CE directives related to safety and electromagnetic compatibility. The report can be consulted on demand. In addition, the Milli-Q® IQ 7003/05/10/15 systems are built using components and practices recommended by UL and has been cUL marked. The registration can be verified on the UL web site: www.ul.com.