MilliporeSigma
All Photos(2)
Key Documents
M3445
Sigma-Aldrich
Magnolol
≥90% (HPLC), from plant
Synonym(s):
2,2′-Bichavicol, 5,5′-Diallyl-2,2′-biphenyldiol
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C18H18O2
CAS Number:
Molecular Weight:
266.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.25
biological source
plant
Quality Level
assay
≥90% (HPLC)
solubility
ethanol: soluble 1 mg/mL
methanol: soluble 1.5 mg/mL
antibiotic activity spectrum
Gram-negative bacteria
Gram-positive bacteria
fungi
application(s)
metabolomics
vitamins, nutraceuticals, and natural products
mode of action
cell membrane | interferes
storage temp.
2-8°C
SMILES string
Oc1ccc(CC=C)cc1-c2cc(CC=C)ccc2O
InChI
1S/C18H18O2/c1-3-5-13-7-9-17(19)15(11-13)16-12-14(6-4-2)8-10-18(16)20/h3-4,7-12,19-20H,1-2,5-6H2
InChI key
VVOAZFWZEDHOOU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Magnolol is a bioactive neolignan extracted from the root and stem bark of different Chinese herbal Magnolia trees. It has a hydroxylated bi-phenoid structure like that of its structural isomer honokiol. It is a lipophilic compound.
Application
Magnolol has been used:
  • to test its effect on muscle atrophy and macrophage subtypes in sarcopenia mouse model
  • to test its inhibitory effect on human umbilical vein endothelial cells (HUVECs) in angiogenesis assay
  • to test its anti-fungal effects on growth, ultrastructure, and biofilm formation of Candida spp. strains
  • in the preparation of immobilized amino-functionalized mesoporous silica particles for prolonged-release studies
Biochem/physiol Actions
Magnolol exerts anti-depressant and, neurodegenerative properties. It is also considered to be a potent anti-cancer agent for its involvement in reducing cell proliferation, cell cytotoxicity, and inducing apoptosis.
Magnolol is a bioactive plant component isolated from the root and stem bark of Magnolia officinalis with antifungal, antibacterial and antioxidant effects. Magnolol also demonstrates anti-inflammatory activity by interferring with NF-κB signaling. It also appears to have anti-inflammatory properties related to vascular disorders such as atherosclerosis due to its ability to inhibit IL-6-induced STAT3 activation.
signalword
Danger
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
ppe
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Hanna Kuk et al.
Scientific reports, 7(1), 17820-17820 (2017-12-21)
Due to gravity the venous vasculature in the lower extremities is exposed to elevated pressure levels which may be amplified by obesity or pregnancy. As a consequence, venules dilate and may be slowly transformed into varicose or spider veins. In
Development of a prolonged-release drug delivery system with magnolol loaded in amino-functionalized mesoporous silica
Stefanache A, et al.,
Applied Sciences, 237-237 (2017)
Jawad Behbehani et al.
Microbial pathogenesis, 113, 209-217 (2017-10-28)
The incidence of oral candidosis has increased in recent years due to the escalation in HIV-infection, cancer treatments, organ transplantation, and diabetes. In addition, corticosteroid use, dentures, and broad-spectrum antibiotic use have also contributed to the problem. Treatment of oral
Abhishek Manoj Ranaware et al.
International journal of molecular sciences, 19(8) (2018-08-15)
The past few decades have witnessed widespread research to challenge carcinogenesis; however, it remains one of the most important health concerns with the worst prognosis and diagnosis. Increasing lines of evidence clearly show that the rate of cancer incidence will
Rajendra Karki et al.
Experimental cell research, 319(20), 3238-3250 (2013-08-03)
Increased proliferation and migration of vascular smooth muscle cells (VSMCs) contribute importantly to the formation of both atherosclerotic and restenotic lesions. The objective of this study was to investigate the effect of magnolol on VSMC migration. The proteolytic activity of
Articles
Chronic inflammation is an underlying factor in the development and progression of many of the chronic diseases of aging, such as arthritis, atherosclerosis, diabetes, and cancer.
Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service