MilliporeSigma
HomeMetabolomics ResearchPapain, Cysteine Protease, Properties & Products
Papain, Cysteine Protease, Properties & Products
E.C. 3.4.22.2
Papaya tree (Carica papaya) with tropical papaya fruits showing a blue sky with clouds through the leaves. Papain is isolated from the papaya latex.
Papaya (Carica papaya)
Physical Properties and Kinetics
Papain is a cysteine protease of the peptidase C1 family. Papain consists of a single polypeptide chain with three disulfide bridges and a sulfhydryl group necessary for activity of the enzyme. 
Molecular weight: 23,406 Da (amino acid sequence)16
Optimal pH for activity: 6.0-7.0
Temperature Optimum for Activity: 65 °C22
pI: 8.75 17; 9.55 18 Spectral properties:
λmax: 278 nm 19
Extinction coefficient, E1% = 25 19
Extinction coefficient, EmM = 57.6 (at 280 nm) 20
Unit Definition: One unit will hydrolyze 1.0 µmole of N-α-benzoyl-L-arginine ethyl ester (BAEE) per minute at pH 6.2 at 25 °C.
Structure of papain enzyme showing the small signal peptide on the end, followed by the larger activation peptide, then various active sites. Disulfides are present around the active sites.
Figure 1.Structure of papain enzyme.
Specificity
Papain will digest most protein substrates more extensively than the pancreatic proteases. Papain exhibits broad specificity, cleaving peptide bonds of basic amino acids, leucine, or glycine. It also hydrolyzes esters and amides. Papain exhibits a preference for an amino acid bearing a large hydrophobic side chain at the P2 position. It does not accept Val at the P1' position.1
Applications
  • Papain is commonly used in cell isolation procedures where it has proven more efficient and less destructive than other proteases on certain tissues. For example, papain has been used to isolate viable, morphologically intact, cortical neurons from postnatal rats.2 Our papain preparation (Product No. P4762) has been used for the isolation of smooth muscle cells.3,4 Papain was found to significantly increase the yield of viable smooth muscle cells while not affecting cell sensitivity to stimulants.5
  • Limited papain digestion has proven useful for structural studies of enzymes and other proteins.6-8
  • Papain is used in red cell serology to modify the red cell surface to enhance or destroy the reactivity of many red cell antigens as an adjunct to grouping, antibody screening, or antibody identification procedures. Papain has also been shown to be useful in platelet serology.9
  • Papain has also been used in the enzymatic synthesis of amino acids, peptides, and other molecules.10-13
  • Fab and F(ab')2 antibody fragments are used in assay systems where the presence of the Fc region may cause problems. In these cases it is preferable to use only the antigen binding (Fab) portion of an antibody. Papain is used routinely for the preparation of Fab fragments from IgG. IgM may also be digested with papain resulting in high yields of homogeneous Fab preparations.15
  • Papain cleaves antibodies into two Fab fragments, which recognize the antigen specifically with their variable region, and one Fc fragment.14 It cleaves above the hinge region containing the disulfide bonds that join the heavy chains, but below the site of the disulfide bond between the light chain and heavy chain. This generates two separate monovalent (containing a single antibody binding site) Fab fragments and an intact Fc fragment. The fragments can be purified by gel filtration, ion exchange, or affinity chromatography. Protocols for antibody digestion and purification of antibody fragments can be found in Antibodies: A Laboratory Manual, E. Harlow and D. Lane, ed., Cold Spring Harbor Laboratory, Cold Spring Harbor, N.Y., 1988.
  • In tissues such as lymph nodes or spleen, or in peripheral blood preparations, cells with Fc receptors (macrophages, monocytes, B lymphocytes, and natural killer cells) are present which can bind the Fc region of intact antibodies, causing background staining in areas that do not contain the target antigen. Use of Fab fragments ensures that the antibodies are binding to the antigen and not to Fc receptors. These fragments may also be desirable for staining cell preparations in the presence of plasma, because they are not able to bind complement, which could lyse the cells. Fab fragments allow more exact localization of the target antigen, i.e. in staining tissue for electron microscopy.
Typical pepsin and papain cleavage of an immunoglobulin G antibody. Generic antibody is on the left with an arrow showing pepsin cleavage above the disulfide bond hinge region into F(ab)2 fragment and Fc fragments. Another arrow shows papain cleavage below the hinge region splitting the antibody into Fab fragments and Fc fragments.
Figure 2.Representation of pepsin and papain cleavage.
Solubility and Solution Stability
Papain is soluble in water at 10 mg/mL. Immediately prior to use, the enzyme is typically diluted in buffer containing ~5 mM L-cysteine. Activation/stabilizing agents include EDTA, cysteine, and dimercaptopropanol.21
Although papain solutions have good temperature stability, the solution stability is pH dependent. Papain solutions are unstable under acidic conditions, i.e., at pH values below 2.8, there is a significant loss in activity. For the active enzyme in solution, the loss in activity is about 1-2% per day, probably as a result of autolysis and/or oxidation.
A common inactive form of papain obtained during isolation is a mixed disulfide formed between the active site sulfhydryl group of the protein and free cysteine.23
Papain solutions are stable to several denaturing agents, i.e., full activity is maintained after recrystallization in 70% methanol and in 8 M urea solutions. However, there is a significant loss in activity when papain is exposed to 10% trichloroacetic acid or to 6 M guanidine hydrochloride.
Related Products
Papain
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Inhibitors
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Substrates
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
GP Moss. [Internet]. EC 3.4.22.2: International Union of Biochemistry and Molecular Biology (IUBMB).[updated 05 Jun 2020; cited 17 Jul 2020]. Available from: https://www.qmul.ac.uk/sbcs/iubmb/enzyme/EC3/4/22/2
2.
Kinoshita K, Sato K, Hori M, Ozaki H, Karaki H. 2003. Decrease in activity of smooth muscle L-type Ca2+ channels and its reversal by NF-?B inhibitors in Crohn's colitis model. American Journal of Physiology-Gastrointestinal and Liver Physiology. 285(3):G483-G493. https://doi.org/10.1152/ajpgi.00038.2003
3.
Driska SP, Laudadio RE, Wolfson MR, Shaffer TH. 1999. A method for isolating adult and neonatal airway smooth muscle cells and measuring shortening velocity. Journal of Applied Physiology. 86(1):427-435. https://doi.org/10.1152/jappl.1999.86.1.427
4.
HASEGAWA M, KOBAYASHI M, OYAMADA H, KAMISHIMA T, YOSHIDA C, OHATA H, MARUYAMA I, MOMOSE K, GOMI Y. 1987. Studies on isolated smooth muscle cells. IX Application of papain for isolation of single smooth muscle cells from guinea-pig taenia coli.. Jpn. J. Smooth Muscle Res.. 23(1):35-46. https://doi.org/10.1540/jsmr1965.23.35
5.
Margossian SS, Lowey S. 1973. Substructure of the myosin molecule. Journal of Molecular Biology. 74(3):301-311. https://doi.org/10.1016/0022-2836(73)90375-6
6.
Margossian SS, Lowey S. 1973. Substructure of the myosin molecule. Journal of Molecular Biology. 74(3):313-330. https://doi.org/10.1016/0022-2836(73)90376-8
7.
Shiozaki K, Yanagida M. 1991. A functional 125-kDa core polypeptide of fission yeast DNA topoisomerase II.. Mol. Cell. Biol.. 11(12):6093-6102. https://doi.org/10.1128/mcb.11.12.6093
8.
Kyer CI. 1995. Information technology law: What does the future hold?. Computer Law & Security Review. 11(3):140-142. https://doi.org/10.1016/s0267-3649(00)80035-7
9.
?-Nitro-?-amino acids as latent ?,?-dehydro-?-amino acid residues in solid-phase peptide synthesis. 2004(10):101. https://doi.org/10.3998/ark.5550190.0005.a11
10.
Rajesh M, Kapila S, Nam P, Forciniti D, Lorbert S, Schasteen C. 2003. Enzymatic Synthesis and Characterization ofl-Methionine and 2-Hydroxy-4-(methylthio)butanoic Acid (HMB) Co-oligomers. J. Agric. Food Chem.. 51(9):2461-2467. https://doi.org/10.1021/jf026093g
11.
Fukuoka T, Tachibana Y, Tonami H, Uyama H, Kobayashi S. 2002. Enzymatic Polymerization of Tyrosine Derivatives. Peroxidase- and Protease-Catalyzed Synthesis of Poly(tyrosine)s with Different Structures. Biomacromolecules. 3(4):768-774. https://doi.org/10.1021/bm020016c
12.
Burton SG, Cowan DA, Woodley JM. 2002. The search for the ideal biocatalyst. Nat Biotechnol. 20(1):37-45. https://doi.org/10.1038/nbt0102-37
13.
Greenfield E. 2014. Antibodies: A Laboratory Manual. 2. New York: Cold Spring Harbor Laboratory Press.
14.
NEWKIRK MM, EDMUNDSON A, WISTAR R, KLAPPER DG, CAPRA JD. 1987. A New Protocol to Digest Human IgM with Papain that Results in Homogeneous Fab Preparations that Can Be Routinely Crystallized. Hybridoma. 6(5):453-460. https://doi.org/10.1089/hyb.1987.6.453
15.
Mitchel R, Chaiken I, Smith E. 1970. The Complete Amino Acid Sequence of Papain. J. Biol. Chem. 2453485-3492.
16.
Smith EL, Kimmel JR, Brown DM. 1954. CRYSTALLINE PAPAIN: II. PHYSICAL STUDIES; THE MERCURY COMPLEX. J. Biol. Chem.. 207533-549.
17.
Sluyterman L, de Graaf M. 1972. The effect of salts upon the pH dependence of the activity of papain and succinyl-papain. Biochimica et Biophysica Acta (BBA) - Enzymology. 258(2):554-561. https://doi.org/10.1016/0005-2744(72)90247-1
18.
Glazer AN, Smith EL. 1961. Phenolic Hydroxyl Ionization in Papain. J. Biol. Chem.. 2362948-51.
19.
Pace CN, Vajdos F, Fee L, Grimsley G, Gray T. 1995. How to measure and predict the molar absorption coefficient of a protein. Protein Sci.. 4(11):2411-2423. https://doi.org/10.1002/pro.5560041120
20.
Arnon R. 1970. [14] Papain.226-244. https://doi.org/10.1016/0076-6879(70)19017-3
21.
Kilara A, Shahani KM, Wagner FW. 1977. Preparation and properties of immobilized papain and lipase. Biotechnol. Bioeng.. 19(11):1703-1714. https://doi.org/10.1002/bit.260191109
22.
Klein IB, Kirsch JF. 1969. The Activation of Papain and the Inhibition of the Active Enzyme by Carbonyl Reagents. J. Biol. Chem.. 2445928-5935.