MilliporeSigma
HomeApplicationsGenomicsAdvanced Gene Editing
Advanced Gene Editing
Genome editing techniques
Targeted genome editing has rapidly become a vital tool across the entire research continuum from early discovery to therapeutic application. By enabling scientists to selectively disrupt, recover, repress, or activate gene expression, modern gene editing methods allow for unprecedented exploration of genetic mechanisms governing biological processes. Application of genome engineering ranges from curing developmental disorders to developing disease-resistant crops.
Featured Categories
The image showcases a close-up view of a multi-well plate being filled with a red liquid by an automated pipetting system. The pipettes are aligned and dispensing the liquid into the wells in unison, with a blurred background emphasizing the precision of the process. This scene is typical in laboratory settings where multiple samples are processed simultaneously for experiments or testing.
CRISPR & Gene Editing
With decades of gene editing experience and an unsurpassed portfolio of CRISPR and ZFN technology patents, Sigma-Aldrich® Advanced Genomics delivers the highest quality reagents for your entire workflow, from early-stage discovery to translational research. Whether you are looking to knockout, knock-in, knockdown, or overexpress your targets, our comprehensive suite of CRISPR gene-editing tools and services will take your research Beyond the Bench.
View Products
X-Tremegene Roche Transfection Reagents Bottles
Transfection Reagents
Transfection reagents, protocols, and resources including Roche X-tremeGENE transfection reagents for DNA, siRNA, miRNA, and CRISPR/Cas9 gene editing components.
Shop Products
Schematic diagram of the CRISPR gene editing system showing the different types of synthetic gRNA for CRISPR.
Schematic diagram of the CRISPR gene editing system showing the different types of synthetic gRNA for CRISPR.
CRISPR – Accurate, Efficient Gene Editing
Gene editing is a specific and targeted change to a DNA sequence and involves the addition, removal or modification of the DNA. The CRISPR-Cas system (evolved in microbes as a defense mechanism) is the basis for a class of gene-editing tools that are enabling advances from health and diagnostics to agriculture and energy. Using CRISPR, researchers have the power to target a specific gene, gene family, or even screen an entire genome.
Sigma-Aldrich® Advanced Genomics offers an industry leading selection of CRISPR-Cas9 proteins to meet your individual research needs. Choose from multiple Cas9 variants (wild-type, enhanced specificity, nickase, GFP-fused, catalytically inactive) and several formats (plasmid, lentivirus, lyophilized protein).
CRISPRi and CRISPRa – Powerful Gene Inhibition and Activation
CRISPRi (CRISPR interference) and CRISPRa (CRISPR activation) deliver highly efficient silencing and activation of genes, respectively, without altering the underlying DNA sequence. When employed in large scale LOF (loss-of-function) and GOF (gain-of-function) screens, researchers are able to identify unique, yet functionally related, gene pathways that are often missed with other methods.
Sigma-Aldrich® Advanced Genomics offers a complete suite of optimized CRISPRi and CRISPRa libraries for gene knockdown and overexpression experiments. Pooled CRISPRi and SAM CRISPRa lentiviral libraries are available off-the-shelf or customized to your specific needs.
Wondering if CRISPR is right for your project? Connect with a Sigma-Aldrich® Advanced Genomics Expert.
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents