Custom & Predesigned RNA Oligos, siRNA Duplexes and Controls
Silencing gene expression has become an important strategy in functional genomics. Bioinformatics software, optimized siRNA reagents, and protocols have now made RNAi experiments fast and convenient. Therefore, RNA interference (RNAi) is a useful technology for research and drug discovery.
We are a leading supplier of custom and predesigned RNA, siRNA, esiRNA, and miRNA as well as transfection reagents for the global life science community.
In addition to the use of cutting-edge technology, competitive prices, and continuous investment in worldwide operations, we also offer:
- Deep scientific expertise and over 35 years of experience in the field - we meet customers’ specifications, no matter how complex
- Outstanding customer service & technical support - our customers are provided with best-in-class service and support
- Our commitment to quality for guaranteed product performance - It is the output of our state-of-the-art analytical instruments, our comprehensive understanding of bioinformatics, coupled with a robust quality management system
Read more about
Custom & Predesigned RNA Oligos and siRNA Duplexes
Positive and Negative siRNA Controls
Product | Product Description | |
---|---|---|
Predesigned siRNA | Extensive collection of human, mouse and rat siRNA. Multiple sequences available, with rankings to indicate performance | Order |
Validated Predesigned siRNA | Validated siRNA (>75% mRNA knockdown) for many common gene targets, suitable for transfection optimization and as positive controls. | Order |
MISSION® Positive Control siRNA | Validated siRNA positive controls used to optimize transfection | Order |
siRNA Duplex in Tubes | Available with a wide range of modifications, purifications and quantities to tailor your unique sequence | Order |
siRNA Duplex in Plates | Supplied in 96 well plates | Order |
Fast siRNA Duplex | Provided in solution at concentrations of 50µM (2 OD) or 100µM (5 OD) - Regionally available US orders must be received by 11 AM CST. European orders must be received by 11 AM CET. | Order |
iScale Oligos - siRNA | Milligram and gram quantities of siRNA suitable for in vivo applications, including target validation and pre-clinical testing | Request Quote |
miRNA | Available with a range of modifications, purifications and quantities to tailor your unique sequence | Order |
RNA Oligos | Single strand oligos containing RNA bases, mixed DNA/RNA or RNA/OMe bases | Order |
Custom & Predesigned esiRNA | Heterogeneous pool of siRNA targeting the same mRNA sequence, resulting in highly specific and effective gene knockdown with lower off-target effects | Order |
SIC001 | Universal Negative Control #1 | Order |
SIC002 | Universal Negative Control #2 | Order |
SIC003 | Fluorescent Universal Negative Control #1 w/Cyanine 3 | Order |
SIC004 | Fluorescent Universal Negative Control #2 w/Cyanine 3 | Order |
SIC005 | Fluorescent Universal Negative Control #1 w/Cyanine 5 | Order |
SIC006 | Fluorescent Universal Negative Control #2 w/Cyanine 5 | Order |
SIC007 | Fluorescent Universal Negative Control #1 w/6-FAM | Order |
SIC008 | Fluorescent Universal Negative Control #2 w/6-FAM | Order |
S1452 | siRNA Transfection Reagent | Order |
Related Product Resources
- Custom Oligos & qPCR Probes Online Tools
Learn more about the tools we offer for oligo and qpcr probe design, including OligoArchitect™ Primer and Probe Design Solutions and OligoEvaluator™: Oligonucleotide Sequence Calculator.
- Custom DNA and RNA Oligos & qPCR Probes Product Resources
Review product and technical resources for our custom predesigned DNA oligos, RNA oligos & qPCR probes including the online tools, case studies, and oligonucleotide label licenses and FAQs such as oligo synthesis, length, modification, and many more.
- Custom Oligo Blanket Purchase Orders
We are pleased to offer a tracking system for your custom oligo Blanket Purchase Orders. Our tracking system provides easy management of expenditures against your Blanket Purchase Order (P.O.). Simply register your Blanket P.O. to take advantage of the FREE tracking.
- RapidRED™ Payment Program
The payment program enables you to minimize paperwork and streamline your online ordering process with only one purchase transaction for setting up and depositing funds into your payment account.
- Oligo Product Reviews by SelectScience®
Custom & oredesigned DNA Oligos peer reviews. Check out what your peers are saying!
Custom & Predesigned RNA Oligos and siRNA Duplexes
MISSION® Predesigned siRNA
Our MISSION® Predesigned siRNA (small interfering RNA) are best-in-class and guarantee gene silencing. We have made ordering predesigned siRNA for single gene targets easy through our online ordering interface. Search your gene of interest by gene name, symbol, RefSeq, or Gene ID number to view the predesigned siRNA sequences available for your gene of interest. Predesigned siRNA sequences are available for human, mouse, and rat species.
Custom siRNA Duplexes
Our high-quality, custom siRNA duplexes are used in gene silencing applications. The siRNAs can be chemically modified to your specifications and are analyzed by mass spectrometry prior to release. Custom siRNA duplexes can be supplied in tube or plate format.
Fast siRNA
For quick synthesis and shipment of custom siRNA opt for our Fast siRNA selection (available in specific regions).
iScale siRNA
If you looking to scale up your experiments and require milligram to gram quantities of custom siRNA duplexes use our iScale online request form.
MISSION® esiRNA
Our MISSION® esiRNA provides RNAi researchers with a proven and simple way to perform post-transcriptional silencing of protein-coding genes and lncRNA (long non-coding RNA).
single-stranded RNA (ssRNA) oligos
We offer standard quantities of single-stranded RNA oligos for various applications. Our custom RNA oligos allow a wide range of modifications, purifications, and quantities tailored to your specific needs.
Positive and Negative siRNA Controls
- MISSION® Positive Control siRNA serves as an ideal complement to any gene silencing experiment. Positive controls are validated siRNA known to achieve high levels (>70%) of knockdown.
- MISSION® siRNA Universal Negative Controls have been used in siRNA transfections and have been tested in vitro in Human, Mouse, and Rat cells.
- MISSION® siRNA Fluorescent Universal Negative Controls are available in two formats Cyanine 3 and Cyanine 5. These are used for easy visualization and assessment of transfection efficiency.
- MISSION® siRNA Fluorescent Universal Negative Control #1, 6-FAM allows easy visualization of the siRNA in your cells by using fluorescence microscopy.
- MISSION® siRNA Transfection Reagent is for the transfection of siRNA to achieve ≥90% silencing efficiency for transient knockdown of eukaryotic gene expression.