Skip to Content
MilliporeSigma
Trifluoroacetic anhydride for GC derivatization, LiChropur™, ≥99.0% (GC)

Trifluoroacetic anhydride

Trifluoroacetic anhydride for GC derivatization, LiChropur™, ≥99.0% (GC)
Synonym(s):
Trifluoroacetic anhydride, TFAA
Linear Formula:
(CF3CO)2O
CAS Number:
Molecular Weight:
210.03
EC Number:

Products

Shipping
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing