
Octyldodecanol

Synonym(s):
2-Octyl-1-dodecanol
Linear Formula:
CH3(CH2)9CH[(CH2)7CH3]CH2OH
CAS Number:
Molecular Weight:
298.55
Products
Shipping
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique