Skip to Content
US
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iron(III) oxide
Synonym(s):
Ferric oxide
Empirical Formula (Hill Notation):
Fe
2
O
3
CAS Number:
1309-37-1
Molecular Weight:
159.69
EC Number:
215-168-2
Products
Shipping
Ships Today (2)
Product Category
salts (1)
Brand
Sigma-Aldrich (4)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Particle Size (µm)
Reagent Type
Core
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
310050
Iron(III) oxide,
powder, <5 μm, ≥96%
Expand
View Pricing
544884
Iron(III) oxide,
nanopowder, <50 nm particle size (BET)
Expand
View Pricing
529311
Iron(III) oxide,
≥99.995% trace metals basis
Expand
View Pricing
15-1345
Iron(III) oxide,
SAJ first grade, ≥98.0%
Expand
View Pricing